Reacti-Vial™ Small Glass Reaction Vials
Thermo Scientific™

Reacti-Vial™ Small Glass Reaction Vials

Use Thermo Scientific™ Reacti-Vial™ Small Reaction Vials in the Reacti-Therm™ Heating and Stirring module.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Color
TS-132211 mLClear
TS-130971 mLAmber
TS-130995 mLAmber
TS-131000.1 mLClear
TS-132200.3 mLClear
TS-132223 mLClear
TS-1322510 mLClear
Catalog number TS-13221
Price (USD)
213.28
Pack of 12
Volume (Metric):
1 mL
Color:
Clear
Price (USD)
213.28
Pack of 12

Use Thermo Scientific™ Reacti-Vial™ Small Reaction Vials in the Reacti-Therm™ Heating and Stirring module. An internal cone makes small sample handling in these vials easy and convenient. The extra thick glass wall magnifies the sample, making these vials ideal for observing chemical reactions. Reacti-Vials are available in a variety of sizes of clear and amber glass, and are supplied with Thermo Scientific™ Open-Top Screw Caps and PTFE/Rubber Discs.

Applications
  • Residue isolation
  • Derivative preparation
  • Maximum sample retrieval
  • Moisture protection
  • Sample storage
  • Precipitations
  • Centrifugations
  • Solvent evaporation

Additional Product Information

  • Supplied 12/pack, complete with Thermo Scientific Open-Top Screw Caps and Thermo Scientific PTFE/Rubber Laminated Discs
  • Available in several sizes
  • Thermo Scientific PTFE/Silicone Discs ordered separately
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
Closure Size13-425
Product TypeVial
SeptumPTFE/Rubber
Thread StyleInternal
Volume (Metric) Total1 mL
Closure TypeScrew Cap (Open-top) with Septum
ColorClear
MaterialGlass
Product LineReacti-Vial
Quantity12 Pk.
Volume (Metric)1 mL
Unit SizePack of 12