Silylation Grade Solvents
Thermo Scientific™
Silylation Grade Solvents
Use specially manufactured and packaged Thermo Scientific™ Silylation Grade Solvents to meet the exacting needs of silylation and other sensitive derivatization reactions.
Catalog NumberStandard Components
TS-20062Acetonitrile
TS-20672Dimethylformamide (DMF)
TS-20684Dimethylsulfoxide (DMSO)
TS-27530Pyridine
Catalog number TS-20062
Price (JPY)
5,280
Online offer
Ends: 31-Mar-2026
8,800
Save 3,520 (40%)
Each
Standard Components:
Acetonitrile
Use specially manufactured and packaged Thermo Scientific™ Silylation Grade Solvents to meet the exacting needs of silylation and other sensitive derivatization reactions. They are purified, dried, and packaged under nitrogen in convenient Thermo Scientific™ Hypo-Vial™ Sample Storage Vials, and are supplied with elastomer septa to enable immediate access to the sample without exposure to moisture and oxygen. Use polar solvents (acetonitrile, dimethylformamide, dimethylsulfoxide, pyridine, tetrahyrdrofuran) to facilitate reactions; nonpolar organic solvents may be used but will not accelerate the rate of reaction.
Recommended Use
Avoid water or alcohol, as TMS reagents react with active hydrogen.
Avoid enolizable ketones.
Use dimethylformamide for steroids and other large molecules.
Use dimethylsulfoxide to prepare TMS derivatives of tertiary alcohols and some compounds with low solubility in other silylation solvents.
Pyridine is an excellent solvent and reaction medium for MS reactions and is an HCI acceptor in reactions involving organochlorosilanes.
Other commonly used solvents include tetrahydrofuran and acetonitrile.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
HazardsFlammable, Harmful, Irritant
Item Description50 mL
Standard ComponentsAcetonitrile
Standard Concentration98 to 100%
Storage4°C
For Use WithGC
Unit SizeEach