BSTFA + 1–10% TMCS Silylation Reagent
Thermo Scientific™

BSTFA + 1–10% TMCS Silylation Reagent

Ideal for difficult-to-silylate compounds, the Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent is derivatizing compounds.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberStandard ComponentsQuantity
TS-38832BSTFA +1% TMCS10 g
TS-38831BSTFA +1% TMCS10 x 1 mL ampules
TS-38833BSTFA +1% TMCS25 g
TS-38834BSTFA +1% TMCS100 g
TS-38840BSTFA +10% TMCS10 x 1 mL Ampules
Catalog number TS-38832
Price (USD)
-
Standard Components:
BSTFA +1% TMCS
Quantity:
10 g

Use Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent for analytes that are difficult to silylate. Stronger than BSTFA alone, this catalyzed formulation is ideal for derivatizing compounds such as fatty acid amides and slightly hindered hydroxyls for GC analysis.

  • Excellent for derivatizing fatty acid amides, slightly hindered hydroxyls and other compounds
  • Catalyzed formulation is stronger than BSTFA alone

For Research Use Only. Not for use in diagnostic procedures.

General References:

  1. Her, G.R., et al. (1985). Anal. Biochem. 151, 292-298.
  2. Shanchun, J., et al. (1994). Org. Geochem. 21, 415-422.
Specifications
Storage2°C to 8°C
For Use WithGC
Quantity10 g
Standard ComponentsBSTFA +1% TMCS
Standard Concentration98 to 100, 0.1 to 1%
Unit SizeEach