Product Image
Thermo Scientific™

Tri-Sil TP (TMSI:Pyridine) Reagent

Derivatize hydroxyls compounds, particularly carbohydrates with Thermo Scientific™ Tri-Sil TP (TMSI:Pyridine) Reagent.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
TS-4923125 mL
TS-4923010 x 1 mL ampules
Catalog number TS-49231
Price (USD)
-
Quantity:
25 mL
Derivatize hydroxyls compounds, particularly carbohydrates with Thermo Scientific™ Tri-Sil TP (TMSI:Pyridine) Reagent. Comprising TMSI in pyridine, this reagent silylates alcohols and phenols, organic acids, hydroxylamines, amino acids, carbohydrates, flavonoids, glycols and polyglycols, nucleotides, steroids, hydroxyl acids, barbituates, narcotics, indoles, and vitamins.
Additonal Product Information

 

Does not react with amines

May be used in the presence of water as long as there is enough reagent present to react with both the water and the sample.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
HazardsFlammable
Storage2°C to 8°C
For Use WithGC
Quantity25 mL
Standard ComponentsPyridine, 1-(Trimethylsilyl)-1 H-imidazole
Standard Concentration65 - 80, 20 - 25%
Unit SizeEach