Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA) Acylation Reagents
Thermo Scientific™

Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA) Acylation Reagents

Prepare volatile and stable fluoracyl derivatives for GC/MS with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA).
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호표준 구성요소수량
TS-63164Heptafluoro-butanoic acid anhydride10 x 1 mL
TS-63163Heptafluoro-butanoic acid anhydride25 g
TS-65191Pentafluoro-propanoic acid anhydride100 g
TS-65193Pentafluoro-propanoic acid anhydride10 x 1 mL
TS-67363Trifluoroacetic acid anhydride100 g
카탈로그 번호 TS-63164
제품 가격(KRW)
-
표준 구성요소:
Heptafluoro-butanoic acid anhydride
수량:
10 x 1 mL
Prepare fluoracyl derivatives for GC/MS and for flame ionization and electron-capture detection techniques with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA). These reagents react readily with alcohols, phenols, and amines.
  • Used to prepare fluoracyl derivatives for GC/MS
  • Produce stable volatile derivatives for FID and ECD techniques
For Research Use Only. Not for use in diagnostic procedures.
사양
표준 농도98 to 100%
스토리지Room Temperature
용도GC/MS
수량10 x 1 mL
표준 구성요소Heptafluoro-butanoic acid anhydride
Unit SizeEach