Pentafluoropropanol Acylation Reagent
Thermo Scientific™

Pentafluoropropanol Acylation Reagent

Improve detection with the Thermo Scientific™ Pentafluoropropanol Acylation Reagent for derivatization.
Have Questions?
Catalog NumberQuantity
TS-6519510 x 1 mL ampules
Catalog number TS-65195
Price (MXN)
-
Quantity:
10 x 1 mL ampules
Enhance electron-capture detection (ECD) and improve GC-MS analysis by using Thermo Scientific™ Pentafluoropropanol Acylation Reagent for derivatization. This acylation reagent reacts readily with alcohols, phenols, and amines to produce perfluoroacyl derivatives which improve ECD response and provide useful structural information for GC-MS applications. Carboxylic acids can be derivatized using a two-step reaction, which involves a reaction with an anhydride followed by a fluorinated alcohol. This derivatization reagent is well suited for confirmation of drugs of abuse.
  • Addition of fluorine atoms into compounds greatly enhances the sensitivity of certain detectors for all those materials
  • Carboxylic acids can be derivatized using a two-step reaction involving reaction with anhydride, followed by a fluorinated alcohol
Specifications
DescriptionPentafluoropropanol
HazardsIrritant
Standard Concentration98 to 100%
StorageRoom Temperature
For Use WithGC/MS
Quantity10 x 1 mL ampules
Standard Components2,2,3,3,3-Pentafluoro-1-propanol
Unit SizeEach