Amies Agar Gel with Charcoal Transport Swabs
Thermo Scientific™

Amies Agar Gel with Charcoal Transport Swabs

Simplify the transportation of throat, vaginal, wound and skin swabs with this Amies Agar Gel Transport Swabs.
Have Questions?
Catalog NumberQuantity
TS0002A50/Pk., 500/Cs.
Catalog number TS0002A
Price (MXN)
-
Quantity:
50/Pk., 500/Cs.

Simplify the transportation of throat, vaginal, wound and skin swabs with Thermo Scientific™ Oxoid™ Amies Agar Gel with Charcoal Transport Swabs. The presence of charcoal in the medium neutralizes bacterial toxins and other inhibitory substances and prolongs the viability of pathogenic organisms. Amies Agar Gel with Charcoal Transport Swabs come with soft rayon tips, which are inert and non-toxic to microorganisms.

Ensure optimal sample collection and recovery using Amies Agar Gel with Charcoal Transport Swabs.

  • Contains media for the maintenance of bacterial sample during transport to the laboratory.
  • Each batch of product is tested for performance using a wide range of aerobic and anaerobic pathogens to ensure adequate recoveries.
  • Lot number/expiry date and tamper-evident tube ensure complete traceability and sterility.
  • “Venturi” Hour-Glass Tube Design prevents disintegration of gel column and eliminates undesirable air pockets, bubbles and breaks in gel which may be harmful to fastidious bacteria.
  • Maximum gel for anerobe preservations at 5mL provides sample viability and protection
  • Product does not contain glass ampules, meaning more safety as there is no need to break any glass and no seals or barriers to puncture or dry out swabs.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionAmies Agar Gel with Charcoal Transport Swabs
Quantity50/Pk., 500/Cs.
Shaft MaterialPlastic
Storage Requirements5°C to 25°C; do not freeze
Tip MaterialRayon
Product TypeTransport Swab
Swab ConfigurationSingle Applicator
Unit SizeEach