TSC Series -86°C Ultra-Low Temperature Chest Freezer, 12.7 cu. ft. (359.6L) capacity, 115V/60Hz, 16A - FAQs
TSC Series -86°C Ultra-Low Temperature Chest Freezer, 12.7 cu. ft. (359.6L) capacity, 115V/60Hz, 16A - FAQs
View additional product information for TSC Series -86°C Ultra-Low Temperature Chest Freezers - FAQs (TSC1390A)