TSC Series -40°C Ultra-Low Temperature Chest Freezer, 17 cu. ft. (481.4L) capacity, 208-230V/60Hz, 6.5A - FAQs
TSC Series -40°C Ultra-Low Temperature Chest Freezer, 17 cu. ft. (481.4L) capacity, 208-230V/60Hz, 6.5A - FAQs
View additional product information for TSC Series -40°C Ultra-Low Temperature Chest Freezers - FAQs (TSC1750D)