TSC Series -86°C Ultra-Low Temperature Chest Freezer, 20 cu. ft. (566.3L) capacity, 115V/60Hz, 16A - FAQs
TSC Series -86°C Ultra-Low Temperature Chest Freezer, 20 cu. ft. (566.3L) capacity, 115V/60Hz, 16A - FAQs
View additional product information for TSC Series -86°C Ultra-Low Temperature Chest Freezers - FAQs (TSC2090A)