TSC Series -86°C Ultra-Low Temperature Chest Freezer, 3 cu. ft. (84.9L) capacity, 115V/60Hz, 10.5A - FAQs
TSC Series -86°C Ultra-Low Temperature Chest Freezer, 3 cu. ft. (84.9L) capacity, 115V/60Hz, 10.5A - FAQs
View additional product information for TSC Series -86°C Ultra-Low Temperature Chest Freezers - FAQs (TSC390A)