CryoMed™ Controlled-Rate Freezer, General Purpose
CryoMed™ Controlled-Rate Freezer, General Purpose
CryoMed™ Controlled-Rate Freezer, General Purpose
CryoMed™ Controlled-Rate Freezer, General Purpose
CryoMed™ Controlled-Rate Freezer, General Purpose
CryoMed™ Controlled-Rate Freezer, General Purpose
Thermo Scientific™

CryoMed™ Controlled-Rate Freezer, General Purpose

Los congeladores de velocidad controlada (CRF) Thermo Scientific CryoMed están diseñados pensando en la fiabilidad, la protección y la trazabilidad de los datos para sus necesidades de preparación de muestras.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCapacidad (métrico)Requisitos eléctricosModelo
TSCM34PV34 L220-230 V, 50/60HzUso general con impresora
TSCM17PA17 L120 V, 60 HzUso general con impresora
TSCM17PL17 L100 V, 50/60 HzUso general con impresora
TSCM17PV17 L220-230 V, 50/60HzUso general con impresora
TSCM34PA34 L120 V, 60 HzUso general con impresora
TSCM34PL34 L100 V, 50/60 HzUso general con impresora
TSCM48PA48 L120 V, 60 HzUso general con impresora
TSCM48PL48 L100 V, 50/60 HzUso general con impresora
TSCM48PV48 L220-230 V, 50/60HzUso general con impresora
TSCM17XA17 L120 V, 60 HzUso general sin impresora
TSCM17XL17 L100 V, 50/60 HzUso general sin impresora
TSCM17XV17 L220-230 V, 50/60HzUso general sin impresora
TSCM34XA34 L120 V, 60 HzUso general sin impresora
TSCM34XL34 L100 V, 50/60 HzUso general sin impresora
TSCM34XV34 L220-230 V, 50/60HzUso general sin impresora
TSCM48XA48 L120 V, 60 HzUso general sin impresora
TSCM48XL48 L100 V, 50/60 HzUso general sin impresora
TSCM48XV48 L220-230 V, 50/60HzUso general sin impresora
Número de catálogo TSCM34PV
Precio (MXN)
-
Capacidad (métrico):
34 L
Requisitos eléctricos:
220-230 V, 50/60Hz
Modelo:
Uso general con impresora
For customers in research, cell and gene therapy, vaccine production, and biobanking, the CryoMed Controlled-Rate Freezer (CRF) provides precise, repeatable freezing results that protect samples from intracellular freezing. The CryoMed CRF features enhanced data traceability via a touch screen user interface and offers customizable freezing profiles while supporting 21 CFR part 11 and GMP needs.

Thermo Scientific CryoMed Controlled-Rate Freezers (CRF) are designed with reliability, protection, and data traceability in mind for your sample preparation and GMP production needs.

User-friendly operation with enhanced data traceability

  • Intuitive touchscreen display allows for easy set-up, operation, and review of a freezing run
  • Six pre-set freezing profiles and space for up to 14 user-defined, 'custom' freeze profiles
  • Integrated touchscreen UI logs usage and events to support 21 CFR part 11 requirements
  • USB data export of .pdf file run logs support 21 CFR part 11
  • User security: 3 levels of user accessibility
  • 'Run last' feature allows the same profile to be run consecutively with the touch of a button
  • PC interface software provides remote operation capabilities, run review, and custom profile creation

Real-time run monitoring for sample protection

  • Chamber and sample temperatures are monitored by Type T thermocouples, eliminating lag time and providing 'real time' responsiveness
  • Standard alarms to alert users of thermocouple failures, heater malfunction, high/low temperature limits, temperature tracking, power failure, and completion of run
  • Remote alarm contacts
  • Optional thermal printer

Reliable temperature performance for high throughput usage

  • Dual solenoid valves are designed to balance LN2 injection volume for precise temperature control and accelerated freezing
  • Preventative maintenance indicator for LN2 solenoid replacement limits downtime
  • Temperature control and uniformity achieved via an air-handling and liquid nitrogen injection ring system
  • Compatible with VHP cleaning techniques
  • Environmentally friendly insulating foam
  • Type 304 stainless steel with exterior powder-coat finish

Warranty varies by region. Check with your local representative for more information

Especificaciones
Amperaje12 A
Material de la cabinaAcero inoxidable
ConectividadCompatible con Thermo Fisher Connect
Salidas de datosExportación de datos a memoria USB
Dimensiones (F x An x Al) exteriores24,3 x 43,3 x 21,7 in (61 x 109,9 x 55,1 cm)
Dimensiones interiores (F x An x Al)13 x 13 x 12 in (33 x 33 x 30,5 cm)
PantallaInterfaz de usuario de pantalla táctil
Requisitos eléctricos220-230 V, 50/60Hz
Acabado exteriorAcero con pintura en polvo
ModeloUso general con impresora
Registro de datos integradoSí
Capacidad de programación del PCSoftware de interfaz de PC compatible
Tipo de tapónUE
Carga de muestrasCarga frontal
TipoCongelador de tasa controlada
Capac. viales<800
GarantíaLa garantía varía según la región. Consulte al representante de ventas local para obtener más información.
Peso (imperial)174 lb
Peso (métrico)78,9 kg
Capacidad (imperial)1,2 pies cúb.
Capacidad (métrico)34 L
Certificaciones/conformidadCE
compresorAir-cooled
Opciones de vigilanciaAlarms, Remote Alarm Contacts
No. of Doors1
ModeloFreestanding
Intervalo de temperaturaDe -180 °C a +50 °C
Tensión208/230 V
Unit SizeEach