CryoMed™ with OPC UA, General Purpose
Thermo Scientific™

CryoMed™ with OPC UA, General Purpose

Thermo Scientific CryoMed Controlled-Rate Freezers with OPC UA are designed with reliability, protection, and data traceability in mind for your sample preparation and GMP production needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)TypeElectrical Requirements
TSCM48FV48 LControlled-rate Freezer with Printer220-230V, 50-60Hz
TSCM34FA34 LControlled-rate Freezer with Printer120V, 50-60Hz
TSCM34FV34 LControlled-rate Freezer with Printer220-230V, 50-60Hz
TSCM34SA34 LControlled-rate Freezer without Printer120V, 50-60Hz
TSCM34SV34 LControlled-rate Freezer without Printer220-230 V, 50-60 Hz
TSCM48FA48 LControlled-rate Freezer with Printer120V, 50-60Hz
TSCM48SA48 LControlled-rate Freezer without Printer120 V, 50-60 Hz
TSCM48SV48 LControlled-rate Freezer without Printer220-230V, 50-60Hz
TSCM17FV17 LControlled-rate Freezer with Printer220-230V, 50-60Hz
TSCM17SA17 LControlled-rate Freezer without Printer120V, 50-60Hz
TSCM17SV17 LControlled-rate Freezer without Printer220-230V, 50-60Hz
TSCM17FA17 LControlled-rate Freezer with Printer120V, 50-60Hz
Catalog number TSCM48FV
Price (USD)
-
Capacity (Metric):
48 L
Type:
Controlled-rate Freezer with Printer
Electrical Requirements:
220-230V, 50-60Hz
For customers in cell and gene therapy and vaccine production, the CryoMed with OPC UA provides precise, repeatable freezing results that protect samples from intracellular freezing. The CryoMed features OPC UA serial communication (ethernet) capabilities, comes standard factory certifications, provides enhanced data traceability via a touch screen user interface and offers customizable freezing profiles while supporting 21 CFR part 11 and GMP needs.

User-friendly operation with enhanced data traceability** – Reduce equipment noncompliance by documenting functionality to support your regulatory requirements with qualification (IQ/OQ) and performance testing services.

Catalog numbers:

Qualification service

  • On-site IQ/OQ service IOQP003507
  • On-site IQ/OQ + temperature mapping IOTQP003507
  • On-site temperature mapping service TEMPMAP

*Available in 23 countries. Please indicate your interest in the 'Request a quote' form and we will come back to you on availability.

**Compliance services are only available in select countries including U.S., Germany, France, U.K., Ireland, Spain, Portugal, Italy, Austria, Switzerland, Norway, Sweden, Denmark, Netherlands. Please state your interest to the 'Request a quote' form.
Specifications
Amperage5.5A
Cabinet MaterialStainless Steel
ConnectivityThermo Fisher Connect compatible
Data OutputsUSB and OPC UA (ethernet)
Dimensions (DxWxH) Exterior24.3 x 49.3 x 21.7 in. (61 x 125.2 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 19 x 12 in. (33 x 48.3 x 30.5 cm)
Electrical Requirements220-230V, 50-60Hz
Exterior FinishPowder-coated Steel
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
Plug TypeCEE 7/7
Sample LoadingFront-loading
TypeControlled-rate Freezer with Printer
Vial Capacity<1185
WarrantyWarranty varies by region. Contact your local sales representative for more information.
Weight (English)191 lb.
Capacity (Metric)48 L
Certifications/ComplianceCE
CompressorAir-cooled
Monitoring OptionsAlarms, Remote Alarm Contacts
No. of Doors1
StyleFreestanding
Temperature Range-180°C to +50°C
Voltage208/230 V
Unit SizeEach