CryoMed™ Controlled-Rate Freezer, General Purpose
CryoMed™ Controlled-Rate Freezer, General Purpose
CryoMed™ Controlled-Rate Freezer, General Purpose
CryoMed™ Controlled-Rate Freezer, General Purpose
CryoMed™ Controlled-Rate Freezer, General Purpose
CryoMed™ Controlled-Rate Freezer, General Purpose
Thermo Scientific™

CryoMed™ Controlled-Rate Freezer, General Purpose

Thermo Scientific CryoMed Controlled-Rate Freezers (CRF) are designed with reliability, protection, and data traceability in mind for your sample preparation needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Electrical RequirementsModel
TSCM48PA48 L120 V, 60HzGeneral Purpose with Printer
TSCM17PA17 L120 V, 60HzGeneral Purpose with Printer
TSCM17PL17 L100 V, 50/60HzGeneral Purpose with Printer
TSCM17PV17 L220-230 V, 50/60HzGeneral Purpose with Printer
TSCM34PA34 L120 V, 60HzGeneral Purpose with Printer
TSCM34PL34 L100 V, 50/60HzGeneral Purpose with Printer
TSCM34PV34 L220-230 V, 50/60HzGeneral Purpose with Printer
TSCM48PL48 L100 V, 50/60HzGeneral Purpose with Printer
TSCM48PV48 L220-230 V, 50/60HzGeneral Purpose with Printer
TSCM17XA17 L120 V, 60HzGeneral Purpose without Printer
TSCM17XL17 L100 V, 50/60HzGeneral Purpose without Printer
TSCM17XV17 L220-230 V, 50/60HzGeneral Purpose without Printer
TSCM34XA34 L120 V, 60HzGeneral Purpose without Printer
TSCM34XL34 L100 V, 50/60HzGeneral Purpose without Printer
TSCM34XV34 L220-230 V, 50/60HzGeneral Purpose without Printer
TSCM48XA48 L120 V, 60HzGeneral Purpose without Printer
TSCM48XL48 L100 V, 50/60HzGeneral Purpose without Printer
TSCM48XV48 L220-230 V, 50/60HzGeneral Purpose without Printer
Catalog number TSCM48PA
Price (USD)
-
Capacity (Metric):
48 L
Electrical Requirements:
120 V, 60Hz
Model:
General Purpose with Printer
For customers in research, cell and gene therapy, vaccine production, and biobanking, the CryoMed Controlled-Rate Freezer (CRF) provides precise, repeatable freezing results that protect samples from intracellular freezing. The CryoMed CRF features enhanced data traceability via a touch screen user interface and offers customizable freezing profiles while supporting 21 CFR part 11 and GMP needs.

Thermo Scientific CryoMed Controlled-Rate Freezers (CRF) are designed with reliability, protection, and data traceability in mind for your sample preparation and GMP production needs.

User-friendly operation with enhanced data traceability

  • Intuitive touchscreen display allows for easy set-up, operation, and review of a freezing run
  • Six pre-set freezing profiles and space for up to 14 user-defined, “custom” freeze profiles
  • Integrated touchscreen UI logs usage and events to support 21 CFR part 11 requirements
  • USB data export of .pdf file run logs support 21 CFR part 11
  • User security: 3 levels of user accessibility
  • “Run last” feature allows the same profile to be run consecutively with the touch of a button
  • PC interface software provides remote operation capabilities, run review, and custom profile creation

Real-time run monitoring for sample protection

  • Chamber and sample temperatures are monitored by Type T thermocouples, eliminating lag time and providing “real time” responsiveness
  • Standard alarms to alert users of thermocouple failures, heater malfunction, high/low temperature limits, temperature tracking, power failure, and completion of run
  • Remote alarm contacts
  • Optional thermal printer

Reliable temperature performance for high throughput usage

  • Dual solenoid valves are designed to balance LN2 injection volume for precise temperature control and accelerated freezing
  • Preventative maintenance indicator for LN2 solenoid replacement limits downtime
  • Temperature control and uniformity achieved via an air-handling and liquid nitrogen injection ring system
  • Compatible with VHP cleaning techniques
  • Environmentally friendly insulating foam
  • Type 304 stainless steel with exterior powder-coat finish

Warranty varies by region. Check with your local representative for more information

Specifications
Amperage12 A
ConnectivityThermo Fisher Connect compatible
Data OutputsUSB data export
Dimensions (DxWxH) Exterior24.3 x 49.3 x 21.7 in. (61 x 125.2 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 19 x 12 in. (33 x 48.3 x 30.5 cm)
DisplayTouchscreen User Interface
Electrical Requirements120 V, 60Hz
Exterior FinishPowder-coated Steel
ModelGeneral Purpose with Printer
On-Board DataloggingYes
PC ProgrammabilityPC Interface Software Supported
Sample LoadingFront-loading
TypeControlled-rate freezer
Vial Capacity<1185
WarrantyWarranty varies by region. Contact your local sales representative for more information
Weight (English)191 lb.
Weight (Metric)86.6 kg
Cabinet MaterialStainless Steel
Capacity (English)1.7 cu. ft.
Capacity (Metric)48 L
Certifications/CompliancecULus
Plug TypeNEMA 5-15 P
Temperature Range-180°C to +50°C
Unit SizeEach