CryoMed™ with OPC UA, Medical Device
Actual product may vary
CryoMed™ with OPC UA, Medical Device
CryoMed™ with OPC UA, Medical Device
CryoMed™ with OPC UA, Medical Device
CryoMed™ with OPC UA, Medical Device
CryoMed™ with OPC UA, Medical Device
Thermo Scientific™

CryoMed™ with OPC UA, Medical Device

Thermo Scientific CryoMed Controlled-Rate Freezers with OPC UA are designed with reliability, protection, and data traceability in mind for your sample preparation and GMP production needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Type
TSCM48TA48 LControlled-rate Freezer with Printer
TSCM34CA34 LControlled-rate Freezer without Printer
TSCM34TA34 LControlled-rate Freezer with Printer
TSCM48CA48 LControlled-rate Freezer without Printer
TSCM17CA17 LControlled-rate Freezer without Printer
TSCM17TA17 LControlled-rate Freezer with Printer
Catalog number TSCM48TA
Price (USD)
-
Capacity (Metric):
48 L
Type:
Controlled-rate Freezer with Printer
For customers in cell and gene therapy and vaccine production, CryoMed with OPC UA provides precise, repeatable freezing results that protect samples from intracellular freezing. The CryoMed features OPC UA serial communication (ethernet) capabilities, comes standard factory certifications, provides enhanced data traceability via a touch screen user interface and offers customizable freezing profiles while supporting 21 CFR part 11 and GMP needs.

User-friendly operation with enhanced data traceability** – Reduce equipment noncompliance by documenting functionality to support your regulatory requirements with qualification (IQ/OQ) and performance testing services.

Catalog numbers:

Qualification service

  • On-site IQ/OQ service IOQP003507
  • On-site IQ/OQ + temperature mapping IOTQP003507
  • On-site temperature mapping service TEMPMAP

*Available in 23 countries. Please indicate your interest in the 'Request a quote' form and we will come back to you on availability.

**Compliance services are only available in select countries including U.S., Germany, France, U.K., Ireland, Spain, Portugal, Italy, Austria, Switzerland, Norway, Sweden, Denmark, Netherlands. Please state your interest to the 'Request a quote' form.

Specifications
Amperage12 A
Cabinet MaterialStainless Steel
ConnectivityThermo Fisher Connect compatible
Data OutputsUSB and OPC UA (ethernet)
Dimensions (DxWxH) Exterior24.3 x 49.3 x 21.7 in. (61 x 125.2 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 19 x 12 in. (33 x 48.3 x 30.5 cm)
DisplayTouchscreen user interface
Electrical Requirements120V, 50-60Hz
Exterior FinishPowder-coated Steel
On-Board DataloggingYes
PC ProgrammabilityPC Interface software supported
Plug TypeNEMA 5-15 P
Sample LoadingFront-loading
TypeControlled-rate Freezer with Printer
Vial Capacity<1185
WarrantyWarranty varies by region. Contact your local sales representative for more information.
Weight (English)191 lb.
Capacity (Metric)48 L
Certifications/CompliancecULus
CompressorAir-cooled
Monitoring OptionsAlarms, Remote Alarm Contacts
No. of Doors1
StyleFreestanding
Temperature Range-180°C to +50°C
Voltage120 V
Unit SizeEach