TSD Series -40°C Upright Ultra-Low Temperature Freezer, 13 cu. ft. (368.1L) capcity, holds 240 boxes, 115V/60Hz, 16A - FAQs
TSD Series -40°C Upright Ultra-Low Temperature Freezer, 13 cu. ft. (368.1L) capcity, holds 240 boxes, 115V/60Hz, 16A - FAQs
View additional product information for TSD Series -40°C Upright Ultra-Low Temperature Freezers - FAQs (TSD40240A)