TSD Series -40°C Upright Ultra-Low Temperature Freezer, 23 cu. ft. (651.3L) capcity, holds 400 boxes, 115V/60Hz, 16A - FAQs
TSD Series -40°C Upright Ultra-Low Temperature Freezer, 23 cu. ft. (651.3L) capcity, holds 400 boxes, 115V/60Hz, 16A - FAQs
View additional product information for TSD Series -40°C Upright Ultra-Low Temperature Freezers - FAQs (TSD40400A)