TSE Series -86°C Upright Ultra-Low Temperature Freezer, 13 cu. ft. (368.1L) capcity, holds 240 boxes, 208-230V/60Hz, 12A, painted galvaneel interior - FAQs
TSE Series -86°C Upright Ultra-Low Temperature Freezer, 13 cu. ft. (368.1L) capcity, holds 240 boxes, 208-230V/60Hz, 12A, painted galvaneel interior - FAQs
View additional product information for TSE Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (TSE240D)