Search
TSE Series -86°C Upright Ultra-Low Temperature Freezer, 17.3 cu. ft. (489.9 L) capcity, holds 320 boxes, 208-230V/60Hz, 12A, painted galvanneal interior
- FAQsView additional product information for TSE Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (TSE320D)
1 product FAQs found