TSE Series -86°C Upright Ultra-Low Temperature Freezer, 23 cu. ft. (651.3 L) capcity, holds 400 boxes, 120V/60Hz, 16A, painted galvanneal interior
- FAQs
TSE Series -86°C Upright Ultra-Low Temperature Freezer, 23 cu. ft. (651.3 L) capcity, holds 400 boxes, 120V/60Hz, 16A, painted galvanneal interior
- FAQs
View additional product information for TSE Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (TSE400A)