TSE Series -86°C Upright Ultra-Low Temperature Freezer, 23 cu. ft. (651.3L) capacity - FAQs
TSE Series -86°C Upright Ultra-Low Temperature Freezer, 23 cu. ft. (651.3L) capacity - FAQs
View additional product information for TSE Series -86°C Upright Ultra-Low Temperature Freezer, 23 cu. ft. (651.3L) capacity - FAQs (TSE400SSD)