TSE Series -86°C Upright Ultra-Low Temperature Freezer, 23 cu. ft. (651.3L) capcity, holds 400 boxes, 230V/50Hz, 12A, painted galvaneel interior - FAQs
TSE Series -86°C Upright Ultra-Low Temperature Freezer, 23 cu. ft. (651.3L) capcity, holds 400 boxes, 230V/50Hz, 12A, painted galvaneel interior - FAQs
View additional product information for TSE Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (TSE400V)