TSG Series Laboratory Freezers
Thermo Scientific™

TSG Series Laboratory Freezers

Green features
TSG Auto-defrost Freezers offer tight temperature performance at -25°C setpoint with a 12-hour controller backup, remote alarm contacts and a 4-year parts and labor warranty. Available in single and double-door configurations, the freezers are sustainably made at a zero waste-to-landfill facility.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCapacidad (imperial)Dimensiones (F x An x Al) exterioresNo. of Doors
TSG2325FA23 pies cúb.39 x 24 x 58 in1
TSG1225FA12 pies cúb.22 x 20 x 52 in1
TSG5025FA50 pies cúb.29 x 53 x 58 in2
Número de catálogo TSG2325FA
Precio (MXN)
-
Capacidad (imperial):
23 pies cúb.
Dimensiones (F x An x Al) exteriores:
39 x 24 x 58 in
No. of Doors:
1
TSG Series Laboratory Freezers are built from the ground up to meet your cold storage needs, offering quiet operation, optimal performance, and sustainable design. The TSG Series is ready to go out of the box, complete with pre-installed casters, onboard alarms, a 12-hour controller backup system, and remote alarm contacts. These auto-defrost freezers also give you peace of mind knowing that your samples are secure with robust temperature performance, all backed by a 4-year parts and labor warranty.

Designed for sample integrity and security

  • Designed to safely store media, reagents, and other laboratory grade storage requiring tight temperature performance
  • Non-invasive adaptive defrost with time and temperature guided cycle
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Standard door locks provide comfort and assurance

Enabling compliance

  • Tested and certified to conform with the design and safety standards of UL 61010, and CE Directive 2011/65/EU and 2015/863

Designed for an ergonomic and comfortable experience

  • Ergonomically designed door handle
  • Easy to clean and adjust epoxy coated shelves
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Self-closing door with 90° stop to assist with inventory loads

Verified sustainability

  • ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life.
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
  • Manufactured in an award-winning, zero-waste to landfill facility

Ready for connectivity and monitoring

  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems

Make it your own

  • Extra and Alternate wire baskets, shelves, and drawers
  • E-Lock Adapter kits
  • 4-20 milliamp transmitters and other remote alarm modules

Services and support

  • Industry Leading 60-month compressor warranty (varies by country, please contact us for details)
  • Optional Validation Testing Programs include field-based temperature mapping and Installation Qualification (IQ) and Operation Qualification (OQ) programs
  • Optional Factory Acceptance Test results including QC Test Log Sheets and QC Temperature Graphs
Especificaciones
Seguridad de accesoYes
Sistemas de apoyoYes
Material de la cabinaAluminum
Capacidad del estante (imperial)100 lb.
Capacidad del estante (métrico)45 kg
Ruedas giratoriasYes
Material de la cámaraAluminio
DescongelaciónAutomático
Dimensiones (F x An x Al) exteriores39 x 24 x 58 in
Modelo de puertaSolid Swing
Requisitos eléctricos115 V/60 Hz
Para utilizar con (aplicación)General Purpose
Frecuencia60 Hz
Características ecológicasENERGY STAR
Registro de datos integradoSí
Tipo de tapónNEMA 5-15 P
RefrigeranteR290
Peso con embalaje (imperial)315 lb.
Peso con embalaje (métrico)143 kg
Resolución de temperatura1°C
Sensor de temperaturaYes
Temperatura (valor de referencia)-25°C
Garantía4 Years (Parts and Labor)
Capacidad (imperial)23 pies cúb.
Capacidad (métrico)700 L
Certificaciones/conformidadcULus, ENERGYSTAR, EPA SNAP, EPA SNAP
compresorBottom-mounted
Opciones de vigilanciaAlarm Module, Battery Back-up
No. of Doors1
Estantes4
ModeloVertical
Intervalo de temperatura-15°C to -29°C
Tensión115 V
Unit SizeEach