TSU™ Series -86°C Upright Ultra-Low Temperature Freezer, 19.4 cu. ft. (547.9L) capacity, holds 400 boxes, 208-230V/60Hz - FAQs
TSU™ Series -86°C Upright Ultra-Low Temperature Freezer, 19.4 cu. ft. (547.9L) capacity, holds 400 boxes, 208-230V/60Hz - FAQs
View additional product information for TSU™ Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (TSU400D)