TSU™ Series -86°C Upright Ultra-Low Temperature Freezer, 28.8 cu. ft. (815.1 L) capacity, holds 600 boxes, 115V/60Hz
- FAQs
TSU™ Series -86°C Upright Ultra-Low Temperature Freezer, 28.8 cu. ft. (815.1 L) capacity, holds 600 boxes, 115V/60Hz
- FAQs
View additional product information for TSU™ Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (TSU600A)