TSU™ Series -86°C Upright Ultra-Low Temperature Freezer, 33.5 cu. ft. (948.7L) capacity, holds 700 boxes, 208-230V/60Hz - FAQs
TSU™ Series -86°C Upright Ultra-Low Temperature Freezer, 33.5 cu. ft. (948.7L) capacity, holds 700 boxes, 208-230V/60Hz - FAQs
View additional product information for TSU™ Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (TSU700D)