TSV Value Lab Refrigerator/Freezer
TSV Value Lab Refrigerator/Freezer
Thermo Scientific™

TSV Value Lab Refrigerator/Freezer

Chill, freeze and retrieve samples safely, simply and economically with the Thermo Scientific™ TSV Value™ Refrigerator/Freezer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English) Refrigerator
TSV07CPSA7.1 cu. ft.
TSV05CPSA5.1 cu. ft.
Catalog number TSV07CPSA
Price (USD)
-
Request A Quote
Capacity (English) Refrigerator:
7.1 cu. ft.
Chill, freeze and retrieve samples safely, simply and economically with the Thermo Scientific™ TSV Value™ Refrigerator/Freezer.
  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally friendly, SNAP compliant hydrocarbon refrigerants and foam
  • Adjustable temperature control
  • Easy to clearn, durable finish
Specifications
Access SecurityNone
Amperage1A
Casters2 adjustable feet in front
ColorWhite
Control TypeMechanical Dial
Dimensions (DxWxH) Exterior21.75 x 21.5 x 56.63 in. (55.2 x 54.6 x 144 cm)
Dimensions (D x W x H) InteriorRefrigerator: 17 x 17.5 x 36.5 in. (43.2 x 44.5 x 92.7 cm)Freezer: 16 x 15.5 x 10.38 in. (40.6 x 39.4 x 26.4 cm)
Door Shelves3
Drawers1
Electrical Requirements115V 60Hz
Energy Usage.39 kWh/Day
For Use With (Application)Indoor general storage of non-critical samples and reagents
Frequency60 Hz
InsulationCyclopentane
InteriorABS Plastic
Min. Clearance12 in. on top; 4 in. sides and rear
ModelTSV07CPSA
RefrigerantR600a hydrocarbon refrigerant
Shipping Weight (Metric)45 kg
TypeGeneral Purpose Combo
Typical Lifetime10 years
Warranty12 months
Capacity (English) Refrigerator7.1 cu. ft.
Capacity (Metric) Refrigerator201 L
CompressorHermetic
DefrostAutomatic (Refrigerator), Manual (Freezer)
Door StyleHinged, Open Left to Right
Exterior FinishEnameled Steel
No. of Doors2
Plug TypeNEMA 5-15 P
Shelves3 Adjustable (Refrigerator); 1 (Freezer)
Temperature Range (Freezer)-12°C to -20°C
Temperature Range (Refrigerator)1°C to 12°C
Voltage115 V
Unit SizeEach