TSV Series 72 cu.ft. Lab Refrigerator
Thermo Scientific™

TSV Series 72 cu.ft. Lab Refrigerator

Green features
Thermo Scientific TSV Series 72 cu.ft. lab refrigerators offer a convenient option for storage of non-critical samples and reagents. Featuring three self-closing glass doors, the 72 cu. ft. TSV Series Lab Refrigerators offer an economical solution for users who need significant space for temperature controlled storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCabinet MaterialExterior Finish
TSV72RSGAStainless SteelStainless Steel
TSV72RPGAPainted SteelPainted Steel
Catalog number TSV72RSGA
Price (USD)
-
Cabinet Material:
Stainless Steel
Exterior Finish:
Stainless Steel
Thermo Scientific TSV Series 72 cu.ft. lab refrigerators offer a convenient option for storage of non-critical samples and reagents. Featuring three self-closing glass doors, the 72 cu. ft. TSV Series Lab Refrigerators offer an economical solution for users who need significant space for temperature controlled storage. Additionally, TSV Series 72 cu.ft. Lab Refrigerators come with remote alarm contacts, high and low temperature alarms, and an adjustbale setpoint with a digital controller.
  • Three self-closing glass doors
  • Remote alarm contacts
  • High and low temperature alarms
  • Digital controller with adjustable temperature setpoint
  • Door ajar alarm
  • Optional stainless steel interior
Specifications
Access SecurityKey lock
Cabinet MaterialStainless Steel
Capacity (English) Shelf100 lb.
Capacity (Metric) Shelf45 kg
CastersYes, customer installed
DefrostAutomatic
Dimensions (DxWxH) Exterior75.7 x 32 x 81 in. (1924 x 813 x 2057 mm)
Dimensions (D x W x H) Interior72 x 28.5 x 61.75 in. (1829 x 724 x 1569 mm)
DisplayDigital
Electrical Requirements120 V/60 Hz
Exterior FinishStainless Steel
Green FeaturesENERGY STAR
Monitoring OptionsSMART VUE Compatible, Remote Alarm Contacts
Ports1 rear access port
RefrigerantR290
Shelves12 Adjustable
Shipping Weight (English)730 lb.
Shipping Weight (Metric)318 kg
Temperature (Setpoint)5°C
TypeValue Enhanced Refrigerator
Warranty2 years parts and labor, 3 additional years compressor parts
Capacity (English)72 cu. ft.
Capacity (Metric)2038 L
Certifications/CompliancecULus
CompressorBottom-mounted
Door StyleGlass Self-closing with Lock
No. of Doors3
Plug TypeNEMA 5-15
StyleFreestanding
Temperature Range4°C to 6°C
Voltage115 V
Unit SizeEach