TSX Series High-Performance Lab Refrigerators
TSX Series High-Performance Lab Refrigerators
TSX Series High-Performance Lab Refrigerators
TSX Series High-Performance Lab Refrigerators
Thermo Scientific™

TSX Series High-Performance Lab Refrigerators

Innovative refrigeration solutions for healthcare and research needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)Door StylePlug TypeElectrical Requirements
TSX2305GV23 cu. ft.GlassCEE 7/7208-230V, 50/60Hz
TSX2305GA23 cu. ft.GlassNEMA 5-15115V, 60Hz
TSX4505GA45.8 cu. ft.Sliding GlassNEMA 5-15115V, 60Hz
TSX4505GD45.8 cu. ft.Sliding GlassNEMA 6-15208-230V, 60Hz
TSX4505GV45.8 cu. ft.Sliding GlassCEE 7/7208-230V, 50Hz
TSX5005GA51.1 cu. ft.Double GlassNEMA 5-15115V, 60Hz
TSX5005GD51.1 cu. ft.Double GlassNEMA 6-15208-230V, 60Hz
TSX5005GV51.1 cu. ft.Double GlassCEE 7/7208-230V, 50Hz
TSX5005SA51.1 cu. ft.Double SolidNEMA 5-15115V, 60Hz
TSX5005SD51.1 cu. ft.Double SolidNEMA 6-15208-230V, 60Hz
TSX5005SV51.1 cu. ft.Double SolidCEE 7/7208-230V, 50Hz
TSX1205GA11.5 cu. ft.GlassNEMA 5-15115V, 60Hz
TSX1205GD11.5 cu. ft.GlassNEMA 6-15208-230V, 60Hz
TSX1205GV11.5 cu. ft.GlassCEE 7/7208-230V, 50Hz
TSX1205SA11.5 cu. ft.SolidNEMA 5-15115V, 60Hz
TSX1205SD11.5 cu. ft.SolidNEMA 6-15208-230V, 60Hz
TSX1205SV11.5 cu. ft.SolidCEE 7/7208-230V, 50Hz
TSX3005GA29.2 cu. ft.GlassNEMA 5-15115V, 60Hz
TSX3005GD29.2 cu. ft.GlassNEMA 6-15208-230V, 60Hz
TSX3005GV29.2 cu. ft.GlassCEE 7/7208-230V, 50Hz
TSX3005SA29.2 cu. ft.SolidNEMA 5-15115V, 60Hz
TSX3005SD29.2 cu. ft.SolidNEMA 6-15208-230V, 60Hz
TSX3005SV29.2 cu. ft.SolidCEE 7/7208-230V, 50Hz
TSX2305GD23 cu. ft.GlassNEMA 6-15208-230V, 50/60Hz
TSX2305SA23 cu. ft.SolidNEMA 5-15115V, 60Hz
TSX2305SD23 cu. ft.SolidNEMA 6-15208-230V, 50/60Hz
TSX2305SV23 cu. ft.SolidCEE 7/7208-230V, 50/60Hz
Catalog number TSX2305GV
Price (USD)
-
Capacity (English):
23 cu. ft.
Door Style:
Glass
Plug Type:
CEE 7/7
Electrical Requirements:
208-230V, 50/60Hz

Thermo Scientific™ TSX Series high-performance lab refrigerators are designed with features that support sample protection and sustainability objectives for the storage of media, reagents, chemotherapy, pharmaceutical and other medical and laboratory-grade storage requiring 3° to 7°C. The Series' V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.

Featuring over 450W of reserve refrigeration capacity, the TSX series has the power to protect your product from temperature swings as a result of frequent and prolonged door access, large product loads, and other activities that introduce hear to the cabinet.

The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.

The drive for sample integrity and security

  • Designed to safely store media, reagents, chemotherapy, pharmaceutical and other medical and laboratory-grade products requiring a temperature range of 3°C to 7°C
  • Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Standard door locks provide comfort and assurance

Enabling compliance

  • Tested to GMP Clean Room Class A / ISO 5 & 6 (ISO EN 14644-1) with easy to clean surfaces support USP <797> and <800> hazardous drug compounding and other clean room environments
  • Tested and certified to conform with the design and safety standards of UL 471, 61010, and CE Directive 2011/65/EU and 2015/863

Designed for an ergonomic and comfortable experience

  • Ergonomically designed door handle with a soft-touch grip
  • The controller is angled to help prevent neck strain
  • Convenient, easy to clean and adjustable epoxy coated full and half shelves
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Bright, LED interior lighting
  • Self-closing multipaned glass or solid doors with 90° stop to assist with inventory loads

Verified sustainability

  • ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life. **
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
  • Manufactured in an award-winning, zero-waste to landfill facility

Ready for connectivity and monitoring

  • Thermo Scientific™ DeviceLink™ Connect ready, enabling you to have a connected experience with your unit so you always—and anywhere— know the safety of your samples and the performance of your refrigerator or freezer.
  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems

Make it your own

  • Alternate Door swing configuration and door types
  • Extra and Alternate wire baskets, shelves, and drawers
  • Free-standing or factory-installed chart recorders
  • Stainless Steel interior finish
  • E-Lock Adapter kits
*Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.
Specifications
Amperage16 A
Cabinet MaterialCold-rolled Steel
Chart RecorderOptional
DefrostAutomatic
Depth (English) Exterior37.8 in.
Depth (English) Interior28.5 in.
Depth (Metric) Exterior96.2 cm
Depth (Metric) Interior72.3 cm
DescriptionTSX Series High-Performance Refrigerator, Glass Door
Dimensions (D x W x H) Interior28.5 x 24 x 58 in. (72.3 x 61 x 147.3 cm)
Dimensions (L x W x H) Exterior37.8 x 28 x 78.6 in. (96.2 x 71.1 x 199.6 cm)
DisplayMicroprocessor
Electrical Requirements208-230V, 50/60Hz
Exterior FinishPainted Steel
Frequency50 Hz
Green FeaturesENERGY STAR
Height (English) Exterior78.6 in.
Height (English) Interior58 in.
Height (Metric) Exterior199.6 cm
Height (Metric) Interior147.3 cm
Monitoring OptionsAlarm, Recorder
Product LineTSX Series
RefrigerantR290
Shelves4
Shipping Weight (English)429 lb.
Shipping Weight (Metric)195 kg
Smart View CompatibilitySmart-Vue Compatible
Temperature (Setpoint)3°-7°C (5°C)
TypeHigh-Performance Refrigerator
Weight (English)364 lb.
Weight (Metric)165 kg
Width (English) Exterior28 in.
Width (English) Interior24 in.
Width (Metric) Exterior71.1 cm
Width (Metric) Interior61 cm
Capacity (English)23 cu. ft.
Capacity (Metric)650 L
Certifications/ComplianceUL, cUL, CE
CompressorV-drive
Door StyleGlass
No. of Doors1
Plug TypeCEE 7/7
StyleUpright
Temperature Range3°C to 7°C
Voltage230 V
Unit SizeEach