TSX Series High-Performance -30°C Auto Defrost Freezers - FAQs
View additional product information for TSX Series High-Performance -30°C Auto Defrost Freezers - FAQs (TSX2330FA, TSX1230FA, TSX1230FD, TSX5030FV, TSX2330FD, TSX5030FD, TSX5030FA, TSX3030FD, TSX3030FA, TSX2330FV, TSX1230FV, TSX3030FV)
2 product FAQs found
What is the temperature range that can be maintained by TSX Series High-Performance -30 degrees C Auto Defrost Freezers?
TSX Series High-Performance -30 degrees C Auto Defrost Freezers can maintain temperatures between -35 and -15 degrees C with a setpoint of -30 degrees C.
Can the TSX Series High-Performance -30 degrees C Auto Defrost Freezers be locked?
Yes, The TSX Series freezers come with standard door locks to offer comfort and assurance, helping ensure the security of the stored materials.
Additionally, the units are equipped with a key-operated triple position switch for main power and alarm, which locks in temperature and alarm setpoints to minimize setpoint error and prevent tampering.
For enhanced access control, E-lock adapter kits are available. These E-locks provide access control with audit-trail capabilities and temperature monitoring. They don't require electricity and are available in both factory- and customer-installed options for various lock systems such as CompX™, Pyxis™, Omnicell™, and McKesson™locks. For additional details, refer to the TSX Series High-Performance Refrigerators and Freezers Brochure.