TSX Universal Series General Purpose Ultra-Low Freezer
TSX Universal Series General Purpose Ultra-Low Freezer
TSX Universal Series General Purpose Ultra-Low Freezer
TSX Universal Series General Purpose Ultra-Low Freezer
TSX Universal Series General Purpose Ultra-Low Freezer
TSX Universal Series General Purpose Ultra-Low Freezer
TSX Universal Series General Purpose Ultra-Low Freezer
TSX Universal Series General Purpose Ultra-Low Freezer
TSX Universal Series General Purpose Ultra-Low Freezer
TSX Universal Series General Purpose Ultra-Low Freezer
Thermo Scientific™

TSX Universal Series General Purpose Ultra-Low Freezer

Green features
TSX Universal Series Ultra-Low Freezers - The next generation of our most trusted ULT platform adapts to your needs, reduces your environmental impact, and is built on our most robust design ever.
製品番号(カタログ番号)モデル保持ラック収容量容量(メートル法)
TSX40086FAGeneral Purpose Ultra-Low Freezer 4002インチボックス(400)16 Racks549 L
TSX50086FAGeneral Purpose Ultra-Low Freezer 5002インチボックス(500)20 Racks682 L
TSX60086FAGeneral Purpose Ultra-Low Freezer 6002インチボックス(600)24 Racks816 L
TSX70086FAGeneral Purpose Ultra-Low Freezer 7002インチボックス(700)28 Racks949 L
TSX40086FA-ULTSUltra-Low Freezer 4002 in. Boxes (400)16 Racks549 L
TSX50086FA-ULTSUltra-Low Freezer 5002 in. Boxes (500)20 Racks682 L
TSX60086FA-ULTSUltra-Low Freezer 6002 in. Boxes (600)24 Racks816 L
TSX70086FA-ULTSUltra-Low Freezer 7002 in. boxes (700)28 Racks950 L
製品番号(カタログ番号) TSX40086FA
価格(JPY)
-
モデル:
General Purpose Ultra-Low Freezer 400
保持:
2インチボックス(400)
ラック収容量:
16 Racks
容量(メートル法):
549 L

TSX Universal—Over 80 years of innovation packed into the next-gen ultra-low freezer. Recognizing the impact of your work, we prioritize sample security and reliability, and our legacy of crafting state-of-the-art lab equipment means our focus remains on user-centric design. The TSX Universal heralds a new era in performance with Universal V-Drive technology, delivering tighter control and faster recovery times. Coupled with an expanded setpoint range, universal voltage, whisper-quiet operation, simplified software updates and much more, TSX Universal seamlessly adapts to your workflow. Redefining sustainability, it is ENERGY STAR† certified with a 32% efficiency improvement, ACT† labeled, and is produced in our Zero-Waste to Landfill† certified facility. TSX Universal ULT—The pinnacle of reliability, sustainability, and innovation.

Reliability at its core:

  • Manufactured excellence: built to the highest standards at our ISO 13485:2016 facilities
  • Proven durability: 10k+ hours of intensive lab and field runtime testing to help ensure long-term reliability
  • Proven quality: each unit undergoes rigorous end-of-line testing for quality control
  • Complete transparency: shipped with factory documentation and a certification of calibration*
  • Peace of mind: backed by our 12-year warranty** for long-term confidence

Built to perform, designed to protect:

  • Enhanced performance with universal V-Drive technology: adapts to user activity to help maintain peak performance and protect what matters most—your samples.
  • Stable temperatures: less than 4°C variation helps keep samples safe anywhere in the cabinet
  • Fast recovery: industry-leading 15-minute door recovery helps minimize temperature fluctuations and downtime
  • Rapid cooling, faster setup: reaches -80°C in under 5 hours—41% faster than previous models
  • Cost effective: uses 33% less energy than previous models , helping reduce costs and environmental impact
  • Quick sample access with active pressure relief: innovative pressure relief technology enables re-entry in just 30 seconds so you can retrieve samples as frequently as needed, helping minimize disruptions
  • Maximum protection & compact footprint: advanced vacuum insulation and environmentally friendly 0 GWP, water-blown foam insulation safeguard samples while providing a compact footprint

Greener by design

  • Less hazardous: uses non-hydrofluorocarbon (HFC) refrigerants and foam insulation which has a 0 GWP and Ozone Depleting Potential (ODP) and is water-blown, helping reduce chemical emissions and outgassing
  • Less waste: contains up to 70% recycled materials, with 90% of waste diverted from landfills at our zero-waste facility***†
  • More energy efficient: ENERGY STAR® certified†, helping reduce lab energy costs and environmental footprint
  • Responsibly packaged: uses non-hydrofluorocarbon (non-HFC) refrigerants with low Global Warming Potential (GWP) and foam insulation, with 0 GWP and Ozone Depleting Potential (ODP), that is water-blown, helping reduce chemical emissions and outgassing
  • Extended life: offers a 12-year warranty, longer than any comparable product on the market

Enhanced experience, optimized for ease

  • Lab-ready convenience: universal voltage, multiple size options, and whisper-quiet operation for easy installation and compatibility
  • Reliable storage and organization: ergonomic handle helps ensure an airtight seal to prevent temperature fluctuations, while adjustable racking maximizes storage efficiency
  • Enhanced security and monitoring: 7 in. UI helps streamline operations, optional HID access enhances security, built-in power and outputs enable real-time monitoring and diagnostics, and an optional lightbar allows instant unit status visibility
  • Easy frost management: 7-point gasket design with an adjustable perimeter heater helps minimize frost build up for easier maintenance
  • Seamless integration: built-in ports for LN2, CO2, and monitoring probes helps ensure compatibility with your lab setup

*Shipped documents:

Certificate of conformance, CE declaration of conformance, certificate of calibration, QC test log sheets, and QC temperature graphs.

**Restrictions and exclusions apply. Warranty is void if the unit is not maintained as set forth in the operation.

***Zero waste defined as 90% of non-hazardous waste diverted from landfill, waste to energy and incineration.

† Information not applicable to TSX Universal ULT freezers with '-ULTS' in the part number (TSX40086FA-ULTS, TSX50086FA-ULTS, TSX60086FA-ULTS.)

仕様
ACT Labelあり
アクセスセキュリティキーロック、南京錠用ループ、セットポイントおよび設定用セキュリティ(オプションのNFCキーカードアクセス)
アンペア数8.9∼4.9 A
バックアップシステムオプションのCO2またはLN2
キャビネット材質傷がつきにくい塗装済み鋼板
キャスター4つの全方向キャスター(ロック付き)
チャンバー材質冷延鋼板
接続性Thermo Scientific™Instrument Connectに対応
霜取りマニュアル
奥行き(ヤードポンド法)外部37.6インチ
奥行き(ヤードポンド法)内部28.3インチ
奥行き(メートル法)外部95.5 cm
奥行き(メートル法)内部71.9 cm
概要TSXユニバーサルシリーズ超低温フリーザー、400ボックス容量、100∼230 V 50/60 Hz
寸法(奥行き☓幅☓高さ)外部37.6x28.3x78 in.(95.5x71.9x198.1 cm)
寸法(奥行き☓幅☓高さ)内部28.3x23.1x51.2 in.(71.9x58.8x130.1 cm)
ディスプレイタッチスクリーンLCD
ドア開放後の回復13分(20°C環境で-75°Cで動作する場合)
ドアスタイル外側(1)、内側(2)
電気的要件100∼230 V 50/60 Hz
エネルギー消費量0.38 kW-hr/day/立方フィート(ENERGY STAR)
外装仕上げ塗装された冷延鋼板
周波数50/60 Hz
グリーン機能ENERGY STAR certified, SNAP Compliant, FGAS Compliant, CA Cooling Act Compliant, Natural Low GWP Refrigerants, Zero Waste to Landfill Manufacturing, Recycled Content and Packaging, ACT Label
保持2インチボックス(400)
絶縁体真空断熱パネル(VIP)+ 水発泡ポリウレタンフォーム(SNAP準拠)
内側ステンレススチールオプション
モデルGeneral Purpose Ultra-Low Freezer 400
内蔵データログ温度、イベント、診断
-80℃でのピーク変動4.7
相シングル
プラグタイプNEMA 5-15
ポート2つの背面アクセスポート
製品ラインTSXユニバーサルシリーズ
ラック収容量16 Racks
冷媒R290(1元目)、R170 + R290混合(2元目)
セットポイントセキュリティフルパスワードまたはPINオプションが付いた2つのモード
Smart View適合性内臓アクセサリ5 VおよびPOEアウトレット
温度分解能0.1°C
温度安定性0.2°C
タイプ超低温フリーザー、縦型
バイアル容量40,000 x 2 mL
電圧補償+-10%
保証保証内容は国により異なります。詳細は当社にお問い合わせください。
水の冷却なし
ワット数コンプレッサー938 W
容量(ヤードポンド法)19.4立方フィート
容量(メートル法)549 L
認証/コンプライアンスcULus, CE, ENERGYSTAR, EPA SNAP
コンプレッサーVドライブ
モニタリングオプションアラーム、レコーダー
No. of Doors1
棚調整可能な3枚の棚
スタイル縦型
温度範囲-40°C∼-86°C
電圧100/230 V
Unit SizeEach