Ultracongeladores de Uso General de la Serie Universal TSX
Ultracongeladores de Uso General de la Serie Universal TSX
Ultracongeladores de Uso General de la Serie Universal TSX
Ultracongeladores de Uso General de la Serie Universal TSX
Ultracongeladores de Uso General de la Serie Universal TSX
Ultracongeladores de Uso General de la Serie Universal TSX
Ultracongeladores de Uso General de la Serie Universal TSX
Ultracongeladores de Uso General de la Serie Universal TSX
Ultracongeladores de Uso General de la Serie Universal TSX
Thermo Scientific™

Ultracongeladores de Uso General de la Serie Universal TSX

Green features
Nuestros congeladores ULT garantizan confiabilidad y seguridad para sus valiosas muestras.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoModeloAdmiteCapacidad de la gradillaCapacidad (métrico)
TSX50086FAGeneral Purpose Ultra-Low Freezer 500Cajas de 2 in. (500)20 Racks682 L
TSX40086FAGeneral Purpose Ultra-Low Freezer 400Cajas de 2 in (400)16 Racks549 L
TSX60086FAGeneral Purpose Ultra-Low Freezer 600Cajas de 2 in. (600)24 Racks816 L
TSX70086FAGeneral Purpose Ultra-Low Freezer 700Cajas de 2 in. (700)28 Racks949 L
TSX70086FA-ULTSUltra-Low Freezer 7002 in. boxes (700)28 Racks950 L
Número de catálogo TSX50086FA
Precio (MXN)
-
Modelo:
General Purpose Ultra-Low Freezer 500
Admite:
Cajas de 2 in. (500)
Capacidad de la gradilla:
20 Racks
Capacidad (métrico):
682 L

TSX Universal—Over 80 years of innovation packed into the next-gen ultra-low freezer. Recognizing the impact of your work, we prioritize sample security and reliability, and our legacy of crafting state-of-the-art lab equipment means our focus remains on user-centric design. The TSX Universal heralds a new era in performance with Universal V-Drive technology, delivering tighter control and faster recovery times. Coupled with an expanded setpoint range, universal voltage, whisper-quiet operation, simplified software updates and much more, TSX Universal seamlessly adapts to your workflow. Redefining sustainability, it is ENERGY STAR certified with a 32% efficiency improvement, ACT labeled, and is produced in our Zero-Waste to Landfill certified facility. TSX Universal ULT—The pinnacle of reliability, sustainability, and innovation.

Reliability at its core:

  • Manufactured excellence: built to the highest standards at our ISO 13485:2016 facilities
  • Proven durability: 10k+ hours of intensive lab and field runtime testing to help ensure long-term reliability
  • Proven quality: each unit undergoes rigorous end-of-line testing for quality control
  • Complete transparency: shipped with factory documentation and a certification of calibration*
  • Peace of mind: backed by our 12-year warranty** for long-term confidence

Built to perform, designed to protect:

  • Enhanced performance with universal V-Drive technology: adapts to user activity to help maintain peak performance and protect what matters most—your samples.
  • Stable temperatures: less than 4°C variation helps keep samples safe anywhere in the cabinet
  • Fast recovery: industry-leading 15-minute door recovery helps minimize temperature fluctuations and downtime
  • Rapid cooling, faster setup: reaches -80°C in under 5 hours—41% faster than previous models
  • Cost effective: uses 33% less energy than previous models , helping reduce costs and environmental impact
  • Quick sample access with active pressure relief: innovative pressure relief technology enables re-entry in just 30 seconds so you can retrieve samples as frequently as needed, helping minimize disruptions
  • Maximum protection & compact footprint: advanced vacuum insulation and environmentally friendly 0 GWP, water-blown foam insulation safeguard samples while providing a compact footprint

Greener by design

  • Less hazardous: uses non-hydrofluorocarbon (HFC) refrigerants and foam insulation which has a 0 GWP and Ozone Depleting Potential (ODP) and is water-blown, helping reduce chemical emissions and outgassing
  • Less waste: contains up to 70% recycled materials, with 90% of waste diverted from landfills at our zero-waste facility***†
  • More energy efficient: ENERGY STAR® certified†, helping reduce lab energy costs and environmental footprint
  • Responsibly packaged: uses non-hydrofluorocarbon (non-HFC) refrigerants with low Global Warming Potential (GWP) and foam insulation, with 0 GWP and Ozone Depleting Potential (ODP), that is water-blown, helping reduce chemical emissions and outgassing
  • Extended life: offers a 12-year warranty, longer than any comparable product on the market

Enhanced experience, optimized for ease

  • Lab-ready convenience: universal voltage, multiple size options, and whisper-quiet operation for easy installation and compatibility
  • Reliable storage and organization: ergonomic handle helps ensure an airtight seal to prevent temperature fluctuations, while adjustable racking maximizes storage efficiency
  • Enhanced security and monitoring: 7 in. UI helps streamline operations, optional HID access enhances security, built-in power and outputs enable real-time monitoring and diagnostics, and an optional lightbar allows instant unit status visibility
  • Easy frost management: 7-point gasket design with an adjustable perimeter heater helps minimize frost build up for easier maintenance
  • Seamless integration: built-in ports for LN2, CO2, and monitoring probes helps ensure compatibility with your lab setup

*Shipped documents:

Certificate of conformance, CE declaration of conformance, certificate of calibration, QC test log sheets, and QC temperature graphs.

**Restrictions and exclusions apply. Warranty is void if the unit is not maintained as set forth in the operation.

***Zero waste defined as 90% of non-hazardous waste diverted from landfill, waste to energy and incineration.

† Information not applicable to TSX Universal ULT freezers with '-ULTS' in the part number (TSX40086FA-ULTS, TSX50086FA-ULTS, TSX60086FA-ULTS.)

Especificaciones
ACT LabelSí
Seguridad de accesoPuertas con llave de seguridad estándar, candado de asa, seguridad en el valor de referencia de temperatura y en la configuración (acceso por tarjeta codificada NFC opcional)
Amperaje8,9-4,9 A
Sistemas de apoyoCO2 o LN2 opcional
Material de la cabinaAcero pintado resistente a los arañazos
Ruedas giratorias4 ruedas giratorias multidireccionales con frenos
Material de la cámaraAcero laminado en frío
ConectividadCompatible con Thermo Scientific™ Instrument Connect
DescongelaciónManual
Fondo exterior (imperial)37,6 in
Fondo interior (imperial)28,3 in
Fondo exterior (métrico)95.5 cm
Fondo interior (métrico)71.9 cm
DescripciónUltracongeladores de la serie TSX Universal, capacidad para 500 cajas, 100/230 V, 50/60 Hz
Dimensiones (F x An x Al) exteriores37,6 x 34,0 x 78 in (95,5 x 86,4 x 198,1 cm)
Dimensiones interiores (F x An x Al)28,3 x 28,8 x 51,2 in (71,9 x 73,0 x 130,1 cm)
PantallaPantalla táctil LCD
Recuperación tras apertura de puertas18 min (funcionamiento desde 20 °C a temperatura ambiente a -75 °C) min
Modelo de puertaExterna (1), interna (2)
Requisitos eléctricos100/230 V, 50/60 Hz
Consumo energético0,31 kw-h/día/ft³ (ENERGY STAR)
Acabado exteriorAcero pintado laminado en frío
Frecuencia50/60 Hz
Características ecológicasMenos peligros, menos residuos, más eficiencia energética, empaquetado responsable y mayor vida útil
AdmiteCajas de 2 in. (500)
AislamientoPaneles de aislamiento de vacío (VIP) + espuma de poliuretano de mezcla de agua (compatible con SNAP)
InteriorOpcional en acero inoxidable
ModeloGeneral Purpose Ultra-Low Freezer 500
Registro de datos integradoTemperatura, eventos, diagnósticos
Variación máxima a -80 ºC3.6
FaseIndividual
Tipo de tapónNEMA 5-15
Puertos2 puertos de acceso posterior
Línea de productosSerie TSX Universal
Capacidad de la gradilla20 Racks
RefrigeranteR290 (primera fase), mezcla R170 + R290 (segunda fase)
Seguridad ajustes control2 modalidades, con opciones de contraseña completa o PIN
Compatibilidad con vista inteligenteSalidas de accesorios integrados 5 V y alimentación a través de Ethernet (POE)
Resolución de temperatura0,1 °C
Estabilidad de la temperatura0,2 °C
TipoUltracongelador, vertical
Capac. viales50,000 x 2 mL
Compensación de voltaje±10%
GarantíaLa garantía varía según el país, póngase en contacto con nosotros para obtener más información
Refrigerado por aguaNo disponible
VatajeCompresor 938 W
Capacidad (imperial)24,1 ft³
Capacidad (métrico)682 L
Certificaciones/conformidadcULus, CE, ENERGYSTAR, EPA SNAP
compresorV-drive
Opciones de vigilanciaAlarma, registrador
No. of Doors1
Estantes3 ajustables
ModeloVertical
Intervalo de temperaturaDe -40 °C a -86 °C
Tensión100/230 V
Unit SizeEach