Stellar Mass Spectrometer eLearning Courses
Thermo Scientific™

Stellar Mass Spectrometer eLearning Courses

The Stellar Mass Spectrometer eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principles and/or user maintenance and troubleshooting.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCourse TopicCourse Level
TTHOFTHFoundational TheoryFoundation
TTHOBOPBasic OperationsRoutine
Catalog number TTHOFTH
Price (USD)
800.00
Each
Course Topic:
Foundational Theory
Course Level:
Foundation
Price (USD)
800.00
Each

The Stellar Mass Spectrometer eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.

The Stellar Mass Spectrometer Foundational Theory eLearning course
Is on the basics of the Thermo Scientific Stellar mass spectrometer. This course provides an introduction to the Stellar instrument, gives an overview of the hardware and basic mass spectrometry principles. This self-paced eLearning program is intended for learners new to mass spectrometry or the Stellar Mass spectrometer.

Some of the topics in this course include:

  • Introduction to the Stellar MS
  • Stellar Hardware
  • Mass Spectrometry fundamentals
  • Ion transmission and detection

The Stellar Mass Spectrometer Basic Operations eLearning course
Will provide the basics of using your instrument. This includes getting your instrument up and running, an overview and introduction to any relevant software. This course will cover getting a detailed introduction to the PRM conductor, as well as provide an introduction and overview to the method editor:

  • PRM Conductor
  • Introduction of method editor
Specifications
Access Availablity30 days upon end user first access to the course
Course TopicFoundational Theory
FormatSelf-paced eLearning
LanguagesEnglish
Course LevelFoundation
Duration60 to 120 min.
For Use With (Equipment)Stellar Mass Spectrometer
Product LineThermo Scientific
Product TypeMass Spectrometry
Target AudienceIntermediate user seeking to benefit from good instrument performance
Training LocationDigital access through Online Learning Management System
Unit SizeEach