TRACE™ 1600 Series GC eLearning courses
Thermo Scientific™

TRACE™ 1600 Series GC eLearning courses

TRACE™ 1600 Series GC eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCourse Topic
TTR16BOPBasic Operations
TTR16FTHFoundational Theory
TTR16MTSUser Maintenance and Troubleshooting
Catalog number TTR16BOP
Price (USD)
490.00
Each
Course Topic:
Basic Operations
Price (USD)
490.00
Each

TRACE™ 1600 Series GC eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.

The TRACE™ 1600 Series GC Foundational Theory eLearning course provides the basics of Gas Chromatography (GC). The uses and applications for gas chromatography are covered as well as some example workflows. This course will also describe the basic components of a GC system and their various functions.

  • GC Applications
  • GC Modules
  • Brief Introduction to Data Processing

This Foundational Theory eLearning course is similar to the Trace™ 1300 Series GC

The TRACE™ 1600 Series GC Basic Operation with Chromeleon™ eLearning course familiarizes users with how to create GC methods, sequences, as well as how to start your run in Chromeleon™ Chromatography Data System (CDS). Instruction on how to process data acquired from your Thermo Scientific™ Trace™ 1300/1600 Series gas chromatography system using Chromeleon Chromatography Data System (CDS) software is also covered. This course is designed for new users who want to learn how to use/run basic experiments on the instrument. Some topics include:

  • Setting parameters through the instrument control pane
  • GC-FID method creation using the SSL injector
  • View of chromatographic data
  • Peak detection and identification

The TRACE™ 1600 Series GC User Maintenance and Troubleshooting eLearning course provides basic troubleshooting and maintenance guidelines on the Thermo Scientific™ TRACE™ 1600 Series gas chromatographs. Its content is broken down into three sections:

  • A brief introduction to the value of timely maintenance and documenting maintenance events.
  • GC troubleshooting: how to approach troubleshooting different areas of concern and how to resolve issues/problems.
  • Basic maintenance procedures on the TRACE Series GC

The content included in this course is also applicable to the Trace™ 1300 Series GC.

Specifications
Access Availablity30 days upon end-user first access to the course
Course TopicBasic Operations
DescriptionTRACE 1600 Series GC basic operations with Chromeleon
FormatOnline Self-paced eLearning
LanguagesEnglish
TechniqueGas Chromatography
Time and DateOn-demand upon course end-user registration confirmation
Course LevelFoundation
Duration<30 min.
For Use With (Equipment)TRACE 1600 Series GC
Product LineThermo Scientific
Product TypeChromatography
Target AudienceNew user seeking training on instrument operation and basic experimentation
Training LocationDigital access through Online Learning Management System
Unit SizeEach

Frequently asked questions (FAQs)

How do I purchase eLearning courses online with a purchase order (PO)?

To purchase an eLearning course with a PO, you need to have a purchasing account on thermofisher.com. For detailed steps on how to register and set up a purchasing account, please review this resource page (https://www.thermofisher.com/us/en/home/support/how-to-register-for-account.html). After your purchasing account has been set up, follow the instructions in this How-To-Guide (https://www.thermofisher.com/us/en/home/products-and-services/services/unity-lab-services/equipment-instrument-education/elearning-how-to-guide.html) for finding and ordering eLearning courses online with a PO.

How do I change to whom an eLearning course for analytical instruments and lab equipment is assigned?

Access Credit Manager to manage the eLearning course assignment. Select the eLearning course order and click on the Assign credits button to remove the existing user assignment.
Note: You can only change the assigned user if they have not yet accessed the eLearning course.

Can I assign an eLearning course for analytical instruments and lab equipment to more than one person?

You can assign an eLearning course for analytical instruments and lab equipment to more than one person only if you purchased multiple quantities. The number of quantities purchased equals the number of users that can be assigned to the course.

How long do I have to assign a purchased eLearning course for analytical instruments and lab equipment to a user?

You have one year from the date of purchase to assign an eLearning course for analytical instruments and lab equipment to a user.

What if the user I am delegating an eLearning course for analytical instruments and lab equipment to does not receive the email on how to manage eLearnings?

Please verify the email address used for delegating. If you are unable to locate the email, you can access Credit Manager directly.