TSQ™ 8000 Duo/Evo Triple Quadrupole GC-MS eLearning courses
Thermo Scientific™

TSQ™ 8000 Duo/Evo Triple Quadrupole GC-MS eLearning courses

TSQ™ 8000 Duo/Evo Triple Quadrupole GC-MS eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCourse Topic
TTS80DFTHFoundational Theory
TTS80DMTSUser Maintenance and Troubleshooting
Catalog number TTS80DFTH
Price (USD)
260.00
Each
Course Topic:
Foundational Theory
Price (USD)
260.00
Each

TSQ™ 8000 Duo/Evo Triple Quadrupole GC-MS eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.

The TSQ™ 8000 Duo/Evo Triple Quadrupole GC-MS foundational theory eLearning course provides a complete understanding of the principles, components, and operation of the triple quadrupole GC-MS instruments. This course explores the theoretical foundations underlying triple quadrupole GC-MS instruments, covering topics such as ionization techniques, SRM (selected ion monitoring) scan type, and data interpretation. Here is a breakdown of the course outline:

  • Introduction to a triple quadrupole GC-MS platform, and the different components involved.
  • Fundamentals of Gas Chromatography, including principles of sample introduction and the separation process
  • Ionization techniques for the triple quadrupole GC-MS:
    • Electron Ionization
    • Chemical Ionization
  • Mass Analysis:
    • Triple quadrupole overview
    • Mechanisms of Collision Induced Dissociation (CID)
  • Operation modes of triple quadrupole MS: Full scan, SIM, and SRM (precursor ion scan and product ion scan)
  • Data interpretation using the instrument software

This Foundational Theory eLearning course is similar and applicable to all triple quadrupole GC-MS systems, including the TSQ™ 8000, TSQ™ 9000, and TSQ™ 9610.

The TSQ™ 8000 (Duo/Evo) Triple Quadrupole GC-MS user maintenance and troubleshooting eLearning course provides basic maintenance and troubleshooting guidelines for Thermo Scientific™ TSQ™ 8000 (Duo/Evo) Triple Quadrupole GC-MS. Topics covered in this course include:

  • Overview of the maintenance consumables for the GC, sampler, and mass spectrometer
  • Recommended maintenance schedules for the autosampler, Trace GC, and TSQ™ 8000 (Duo/Evo) mass spectrometer
  • Instructions on basic maintenance tasks and troubleshooting guidelines.

The content included in this course also applies to the TSQ™ 8000, TSQ™ 9000, and TSQ™ 9610 Triple Quadrupole GC-MS systems.

Specifications
Access Availablity30 days upon end-user first access to the course
Course TopicFoundational Theory
DescriptionTSQ 8000 Duo/Evo Triple Quadrupole GC-MS foundational theory
FormatOnline Self-paced eLearning
LanguagesEnglish
TechniqueGas Chromatography Mass Spectrometry
Time and DateOn-demand upon course end-user registration confirmation
Course LevelFoundation
Duration60 to 120 min.
For Use With (Equipment)TSQ 8000 Duo/Evo Triple Quadrupole GC-MS
Product LineThermo Scientific
Product TypeMass Spectrometry
Target AudienceNew user seeking training on instrument operation and basic experimentation
Training LocationDigital access through Online Learning Management System
Unit SizeEach

Frequently asked questions (FAQs)

How do I purchase eLearning courses online with a purchase order (PO)?

To purchase an eLearning course with a PO, you need to have a purchasing account on thermofisher.com. For detailed steps on how to register and set up a purchasing account, please review this resource page (https://www.thermofisher.com/us/en/home/support/how-to-register-for-account.html). After your purchasing account has been set up, follow the instructions in this How-To-Guide (https://www.thermofisher.com/us/en/home/products-and-services/services/unity-lab-services/equipment-instrument-education/elearning-how-to-guide.html) for finding and ordering eLearning courses online with a PO.

How do I change to whom an eLearning course for analytical instruments and lab equipment is assigned?

Access Credit Manager to manage the eLearning course assignment. Select the eLearning course order and click on the Assign credits button to remove the existing user assignment.
Note: You can only change the assigned user if they have not yet accessed the eLearning course.

Can I assign an eLearning course for analytical instruments and lab equipment to more than one person?

You can assign an eLearning course for analytical instruments and lab equipment to more than one person only if you purchased multiple quantities. The number of quantities purchased equals the number of users that can be assigned to the course.

How long do I have to assign a purchased eLearning course for analytical instruments and lab equipment to a user?

You have one year from the date of purchase to assign an eLearning course for analytical instruments and lab equipment to a user.

What if the user I am delegating an eLearning course for analytical instruments and lab equipment to does not receive the email on how to manage eLearnings?

Please verify the email address used for delegating. If you are unable to locate the email, you can access Credit Manager directly.