TSQ™ 8000 Triple Quadrupole GC-MS eLearning Courses
Thermo Scientific™

TSQ™ 8000 Triple Quadrupole GC-MS eLearning Courses

TSQ™ 8000 Triple Quadrupole GC-MS eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCourse Topic
TTS80MTSUser Maintenance and Troubleshooting
TTS80FTHFoundational Theory
TTS80DBOPBasic Operations
TTS80BOPIntermediate Level Operations
Catalog number TTS80MTS
Price (USD)
775.00
Each
Course Topic:
User Maintenance and Troubleshooting
Price (USD)
775.00
Each

TSQ™ 8000 Triple Quadrupole GC-MS eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.

The TSQ™ 8000 Triple Quadrupole GC-MS foundational theory eLearning course provides a complete understanding of the principles, components, and operation of the triple quadrupole GC-MS instruments. This course explores the theoretical foundations underlying triple quadrupole GC-MS instruments, covering topics such as ionization techniques, SRM (selected ion monitoring) scan type, and data interpretation. Here is a breakdown of the course outline:

  • Introduction to a triple quadrupole GC-MS platform, and the different components involved.
  • Fundamentals of Gas Chromatography, including principles of sample introduction and the separation process
  • Ionization techniques for the triple quadrupole GC-MS:
    • Electron Ionization
    • Chemical Ionization
  • Mass Analysis:
    • Triple quadrupole overview
    • Mechanisms of Collision Induced Dissociation (CID)
  • Operation modes of triple quadrupole MS: Full scan, SIM, and SRM (precursor ion scan and product ion scan)
  • Data interpretation using the instrument software

This Foundational Theory eLearning course is similar and applicable to all triple quadrupole GC-MS systems, including the TSQ™ 8000 (Duo/Evo), TSQ™ 9000, and TSQ™ 9610.

The TSQ™ 8000 Triple Quadrupole GC-MS user maintenance and troubleshooting eLearning course provides basic maintenance and troubleshooting guidelines for Thermo Scientific™ TSQ™ 8000 Triple Quadrupole GC-MS. Topics covered in this course include:

  • Overview of the maintenance consumables for the GC, sampler, and mass spectrometer
  • Recommended maintenance schedules for the autosampler, Trace GC, and TSQ™ 8000 mass spectrometer
  • Instructions on basic maintenance tasks and troubleshooting guidelines.

The content included in this course also applies to the TSQ™ 8000 (Duo/Evo), TSQ™ 9000, and TSQ™ 9610 Triple Quadrupole GC-MS systems.

Specifications
Access Availablity30 days upon end-user first access to the course
Course TopicUser Maintenance and Troubleshooting
FormatSelf-paced eLearning
LanguagesEnglish
TechniqueGas Chromatography Mass Spectrometry
Time and DateOn-demand upon course end-user registration confirmation
Course LevelRoutine
Duration30 to 60 min.
For Use With (Equipment)TSQ 8000 Triple Quadrupole GC-MS
Product LineThermo Scientific
Product TypeMass Spectrometry
Target AudienceIntermediate user seeking to benefit from good instrument performance
Training LocationDigital access through Online Learning Management System
Unit SizeEach