TSQ™ 9610 Triple Quadrupole GC-MS Basic Operations
Thermo Scientific™

TSQ™ 9610 Triple Quadrupole GC-MS Basic Operations

This eLearning course discusses basic operational guidelines providing users with instructions on how to operate and tune their GC-MS system.
Have Questions?
Catalog NumberDuration
TTS96BOP60 to 120 min.
Catalog number TTS96BOP
Price (USD)
490.00
Each
Duration:
60 to 120 min.
Price (USD)
490.00
Each

The TSQ™ 9610 Triple Quadrupole GC-MS Basic Operations eLearning course provides operational guidelines for Thermo Scientific™ TSQ™ 9610 Triple Quadrupole GC-MS.

The TSQ™ 9610 Triple Quadrupole GC-MS Basic Operations eLearning course provides operational guidelines for Thermo Scientific™ TSQ™ 9000 Triple Quadrupole GC-MS, including:

  • An overview of the basic operation of the autosampler and TRACE™ GC series
  • Basic user operational guidelines of the TSQ™ series
  • How to operate the GC-MS system using Chromeleon™ CDS
  • An introduction to the different TSQ GCMS tuning options

The content included in this course also applies to the TSQ™ 8000 (Duo/Evo), TSQ™ 8000, and TSQ™ 9000 Triple Quadrupole GC-MS systems.

Specifications
Access Availablity30 days upon end-user first access to the course
Course TopicBasic Operations
DescriptionTSQ 9610 Triple Quadrupole GC-MS basic operations
FormatSelf-paced eLearning
LanguagesEnglish
Course LevelRoutine
Duration60 to 120 min.
For Use With (Equipment)TSQ 9610 Triple Quadrupole GC-MS
Product LineThermo Scientific
Product TypeMass Spectrometry
Target AudienceIntermediate user seeking to benefit from good instrument performance
Training LocationDigital access through Online Learning Management System
Unit SizeEach