TSQ Endura™ Triple Quadrupole MS eLearning Courses
Thermo Scientific™

TSQ Endura™ Triple Quadrupole MS eLearning Courses

TSQ Endura™ Triple Quadrupole MS eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCourse Topic
TTSQENFTHFoundational Theory
TTSQENBOPBasic Operations
Catalog number TTSQENFTH
Price (USD)
260.00
Each
Course Topic:
Foundational Theory
Price (USD)
260.00
Each
TSQ Endura™ Triple Quadrupole MS eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.

The TSQ Endura™ Triple Quadrupole MS foundational theory eLearning course introduces how triple-stage quadrupole mass spectrometers work, the basic hardware configuration of TSQ Series II systems, and the different scan types they can perform.

Some of the concepts and topics presented include:

  • Principles of MS/MS operation
  • Specificity and sensitivity of mass spectrometers
  • Basic hardware configuration
  • Overview of the vacuum system
  • Components inside the mass spectrometer
  • Scan types

This Foundational Theory eLearning course is similar and applicable to all TSQ Series II instruments; e.g. TSQ Quantiva™, TSQ Altis™, TSQ Quantis™, TSQ Fortis™, TSQ Altis™ Plus, TSQ Quantis™ Plus, and TSQ Fortis™ Plus.

The TSQ Endura™ Triple Quadrupole MS Basic Operations eLearning course guides the basic principles of operation of the instrument.

This course includes:

  • A series of video and particulate animations explaining the basics of the following scan types:
    • SIM (Selected Ion Monitoring)
    • Full Scan
    • Selectivity by fragmentation
    • Product ion scan
    • SRM (Selected Reaction Monitoring)
    • Precursor ion scan
    • Neutral Loss Scan
  • Video demonstrations of procedures to calibrate the instrument, including hardware setup, source positioning, review of calibration status, system preparation and priming, tips, and parameters to consider while evaluating spray stability, and running a system check or system calibration.

This eLearning course is similar and applicable to all TSQ Series II instruments; e.g. TSQ Quantiva™, TSQ Altis™, TSQ Quantis™, TSQ Fortis™, TSQ Altis™ Plus, TSQ Quantis™ Plus, and TSQ Fortis™ Plus.

TheTSQ Endura™ Triple Quadrupole MS Basic Operations eLearning course guides the basic principles of operation of the instrument.

This course includes:

  • A series of video & particulate animations explaining the basics of the following scan types:
    • SIM (Selected Ion Monitoring)
    • Full Scan
    • Selectivity by fragmentation
    • Product ion scan
    • SRM (Selected Reaction Monitoring)
    • Precursor ion scan
    • Neutral Loss Scan
  • Video demonstrations of procedures to calibrate the instrument, including hardware setup, source positioning, review of calibration status, system preparation and priming, tips, and parameters to consider while evaluating spray stability, and running a system check or system calibration.

This eLearning course is similar and applicable to all TSQ Series II instruments; e.g. TSQ Quantiva™, TSQ Altis™, TSQ Quantis™, TSQ Fortis™, TSQ Altis™ Plus, TSQ Quantis™ Plus, and TSQ Fortis™ Plus.

Specifications
Access Availablity1 month upon end-user first access to the course
Course TopicFoundational Theory
FormatOnline Self-paced eLearning
LanguagesEnglish
TechniqueLiquid Chromatography Mass Spectrometry
Time and DateOn-demand upon course end-user registration confirmation
Course LevelFoundation
Duration60 to 120 min.
For Use With (Equipment)TSQ Endura Triple Quadrupole MS
Product LineThermo Scientific
Product TypeMass Spectrometry
Target AudienceNew user seeking training on instrument operation and basic experimentation
Training LocationDigital access through Online Learning Management System
Unit SizeEach

Frequently asked questions (FAQs)

How do I change to whom an eLearning course for analytical instruments and lab equipment is assigned?

Access Credit Manager to manage the eLearning course assignment. Select the eLearning course order and click on the Assign credits button to remove the existing user assignment.
Note: You can only change the assigned user if they have not yet accessed the eLearning course.

Can I assign an eLearning course for analytical instruments and lab equipment to more than one person?

You can assign an eLearning course for analytical instruments and lab equipment to more than one person only if you purchased multiple quantities. The number of quantities purchased equals the number of users that can be assigned to the course.

How long do I have to assign a purchased eLearning course for analytical instruments and lab equipment to a user?

You have one year from the date of purchase to assign an eLearning course for analytical instruments and lab equipment to a user.

What if the user I am delegating an eLearning course for analytical instruments and lab equipment to does not receive the email on how to manage eLearnings?

Please verify the email address used for delegating. If you are unable to locate the email, you can access Credit Manager directly.

What if the user I am delegating an eLearning course for analytical instruments and lab equipment to does not have a thermofisher.com account?

If the user you have assigned administrative rights to does not have a thermofisher.com account, they will have to register for an account on thermofisher.com to assign the eLearning course. This account does not need to be a verified account and can be created by entering the following information:

- First Name
- Last Name
- Email address
- Password

The email address used to create the account will need to be the same email used to delegate the administration of the eLearning course.