Kligler Iron Slant Agar With Urea
Thermo Scientific™

Kligler Iron Slant Agar With Urea

Differentiate enteric bacteria based on their ability to ferment glucose and lactose, produce hydrogen sulfide (H2S), and hydrolyze urea, aiding in their identification.
Have Questions?
Número de catálogoPeso de llenado
TV5004DDe 8,5 a 9,5 g
Número de catálogo TV5004D
Precio (EUR)
-
Peso de llenado:
De 8,5 a 9,5 g
Kligler Iron Slant Agar With Urea is used to differentiate enteric bacteria based on their ability to ferment glucose and lactose, produce hydrogen sulfide (H2S), and hydrolyze urea, aiding in their identification.
Especificaciones
ColorRed Orange, Translucent
Peso de llenadoDe 8,5 a 9,5 g
FormatoPoured Tube, Slant Agar
Tipo de empaquetado50 Tubes/Box
Cantidad50/Pk.
Duración de almacenamiento14 Weeks
Origen de fabricaciónGermany
Tipo de productoAgar
Unit SizeEach
Contenido y almacenamiento
2°C to 25°C