Phosphate Buffered Saline (PBS) (Gamma?Irradiated)
Thermo Scientific™

Phosphate Buffered Saline (PBS) (Gamma?Irradiated)

A sterile, isotonic diluent or rinse solution for sample preparation, microbial dilution, or washing in microbiological and laboratory procedures.
Have Questions?
Número de catálogoPeso de llenado
TV5050BDe 8,5 a 9,5 g
Número de catálogo TV5050B
Precio (MXN)
-
Peso de llenado:
De 8,5 a 9,5 g
Phosphate Buffered Saline (PBS) (Gamma?Irradiated) is a sterile, isotonic diluent or rinse solution for sample preparation, microbial dilution, or washing in microbiological and laboratory procedures.
Especificaciones
ColorColorless, Transparent
Peso de llenadoDe 8,5 a 9,5 g
FormatoPoured Tube
Tipo de empaquetado50 Tubes/Box
Cantidad50/Pk.
Duración de almacenamiento52 Weeks
Origen de fabricaciónGermany
Tipo de productoMicrobiology Media
Unit SizeEach
Contenido y almacenamiento
2°C to 25°C