Lauryl Tryptose Broth with MUG and Tryptophan (with Durham Tube)
Thermo Scientific™

Lauryl Tryptose Broth with MUG and Tryptophan (with Durham Tube)

Enumerate presumptive Escherichia coli as well as other coliforms, using the most portable number (MPN) method using this chromogenic media.
Have Questions?
Catalog NumberFill Weight
TV5079E9.5 to 10.5 g
Catalog number TV5079E
Price (USD)
-
Fill Weight:
9.5 to 10.5 g
Enumerate presumptive Escherichia coli as well as other coliforms, using the most portable number (MPN) method using Lauryl Tryptose Broth with MUG and Tryptophan (with Durham Tube).
Specifications
ColorMaize Yellow, Transparent
Fill Weight9.5 to 10.5 g
FormatPoured Tube
Packaging Type50 Tubes/Box
Quantity50/Pk.
Shelf Life52 Weeks
Manufacturing OriginGermany
Product TypeMicrobiology Media
Unit SizeEach
Contents & Storage
2°C to 12°C