Vanquish™ Charged Aerosol Detectors Series eLearning courses
Thermo Scientific™

Vanquish™ Charged Aerosol Detectors Series eLearning courses

Vanquish™ Charged Aerosol Detectors Series eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCourse Topic
TVCADFTHFoundational Theory
TVCADBOPBasic Operations
Catalog number TVCADFTH
Price (USD)
75.00
Each
Course Topic:
Foundational Theory
Price (USD)
75.00
Each

Vanquish™ Charged Aerosol Detectors Series eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.

The Vanquish™ Charged Aerosol Detectors Series Foundational Theory eLearning course will provide a basic overview of fundamentals of Charged Aerosol Detection (CAD). It will also cover use of the power function and robust calibration practices to achieve the best quantitative results. Some modules of the course include:

  • An overview of fundamentals of Charged Aerosol Detection (CAD)
  • A review of principles of operation and its complimentary features
  • An overview of CAD response characteristics
  • Description of recommended calibration practices
  • How to determine an optimal Power Function Value (PFV)

This Foundational Theory eLearning course is similar to the Corona™ Veo™ Series.

The Vanquish™ Charged Aerosol Detectors Series Basic Operations with Chromeleon™ eLearning course provides basic overview, operation and routine maintenance guidelines for the Thermo Scientific™ Dionex™ Corona™ Veo™ and Vanquish™ Charged Aerosol Detectors (CAD). It will also cover how to run an inverse gradient on the Charged Aerosol Detector (CAD). This course is designed for new users who want to learn how to use/run basic experiments on the instrument. Some modules of the course include:

  • Overview of each detector: Corona™ Veo™ CAD & Vanquish™ CAD
  • Basic operational tips
  • Routine (daily, regularly, annually) maintenance, including video demonstrations on the removal, cleaning, and re-installation of the nebulizer.

This Basic Operations eLearning course is similar to the Corona™ Veo™ Series.

Specifications
Course TopicFoundational Theory
DescriptionVanquish Charged Aerosol Detectors Series foundational theory
FormatOnline Self-paced eLearning
TechniqueLiquid Chromatography
Time and DateOn-demand upon course end-user registration confirmation
Course LevelFoundation
Duration<30 min.
For Use With (Equipment)Vanquish Charged Aerosol Detector
Product LineThermo Scientific
Product TypeChromatography
Target AudienceNew user seeking training on instrument operation and basic experimentation
Training LocationDigital access through Online Learning Management System
Unit SizeEach

Frequently asked questions (FAQs)

How do I purchase eLearning courses online with a purchase order (PO)?

To purchase an eLearning course with a PO, you need to have a purchasing account on thermofisher.com. For detailed steps on how to register and set up a purchasing account, please review this resource page (https://www.thermofisher.com/us/en/home/support/how-to-register-for-account.html). After your purchasing account has been set up, follow the instructions in this How-To-Guide (https://www.thermofisher.com/us/en/home/products-and-services/services/unity-lab-services/equipment-instrument-education/elearning-how-to-guide.html) for finding and ordering eLearning courses online with a PO.

How do I change to whom an eLearning course for analytical instruments and lab equipment is assigned?

Access Credit Manager to manage the eLearning course assignment. Select the eLearning course order and click on the Assign credits button to remove the existing user assignment.
Note: You can only change the assigned user if they have not yet accessed the eLearning course.

Can I assign an eLearning course for analytical instruments and lab equipment to more than one person?

You can assign an eLearning course for analytical instruments and lab equipment to more than one person only if you purchased multiple quantities. The number of quantities purchased equals the number of users that can be assigned to the course.

How long do I have to assign a purchased eLearning course for analytical instruments and lab equipment to a user?

You have one year from the date of purchase to assign an eLearning course for analytical instruments and lab equipment to a user.

What if the user I am delegating an eLearning course for analytical instruments and lab equipment to does not receive the email on how to manage eLearnings?

Please verify the email address used for delegating. If you are unable to locate the email, you can access Credit Manager directly.