Vanquish™ Horizon UHPLC System eLearning courses
Thermo Scientific™

Vanquish™ Horizon UHPLC System eLearning courses

The Vanquish™ Horizon UHPLC System eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCourse Topic
TVQHFTHFoundational Theory
TVQHBOP
Catalog number TVQHFTH
Price (USD)
250.00
Each
Course Topic:
Foundational Theory
Price (USD)
250.00
Each

The Vanquish™ Horizon UHPLC System eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.

The Vanquish™ Horizon UHPLC System Foundational Theory eLearning course is an introduction to the general theories associated with High Performance Liquid Chromatography (HPLC). The course examines basic chromatography theories and how different factors affect the chromatography of different types of samples. The standard parts of the HPLC system and analysis of samples will be explored. Some of the modules in this course include:

  • Identifying key components of a chromatogram and the components that cause variations
  • Different separation types based on types of columns, packing materials and flow rates
  • Basic HPLC modules and their functions
  • Basic examples of separation and the influence of altering chromatography conditions.

This Foundational Theory eLearning course is similar to the Vanquish™ Horizon, Vanquish™ Core and the UltiMate™ 3000.

The Vanquish™ Horizon UHPLC System Basic Operations with Chromeleon™ eLearning course will begin by teaching general HPLC considerations that are important to understand before using your instrument. Next, you will complete a series of eLearning modules on each of the instrument components, including various types of pumps and detectors available with your instrument. You will conclude the course with training on how to equilibrate your instrument and the specifics of setting up automated analysis using Thermo Scientific™ Chromeleon™ Chromatography Data System (CDS) software. This course is designed for new users who want to learn how to use/run basic experiments on the instrument. Included in the eLearning series are the following learning modules:

  • General HPLC considerations
  • Pump operating principles and key parameters
  • Autosampler operating principles and key parameters
  • Column compartment operating principles and key parameters
  • Detector operating principles and key parameters
  • Instrument equilibration
  • Automated analysis

This Basic Operations eLearning course is similar to the Vanquish™ Flex and Vanquish™ Core.

Specifications
Access Availablity30 days upon end-user first access to the course
Course TopicFoundational Theory
DescriptionVanquish Horizon UHPLC System foundational theory
FormatOnline Self-paced eLearning
LanguagesEnglish
TechniqueLiquid Chromatography
Time and DateOn-demand upon course end-user registration confirmation
Course LevelFoundation
Duration60 to 120 min.
For Use With (Equipment)Vanquish Horizon UHPLC System
Product LineThermo Scientific
Product TypeChromatography
Target AudienceNew user seeking training on instrument operation and basic experimentation
Training LocationDigital access through Online Learning Management System
Unit SizeEach

Frequently asked questions (FAQs)

How do I purchase eLearning courses online with a purchase order (PO)?

To purchase an eLearning course with a PO, you need to have a purchasing account on thermofisher.com. For detailed steps on how to register and set up a purchasing account, please review this resource page (https://www.thermofisher.com/us/en/home/support/how-to-register-for-account.html). After your purchasing account has been set up, follow the instructions in this How-To-Guide (https://www.thermofisher.com/us/en/home/products-and-services/services/unity-lab-services/equipment-instrument-education/elearning-how-to-guide.html) for finding and ordering eLearning courses online with a PO.

How do I change to whom an eLearning course for analytical instruments and lab equipment is assigned?

Access Credit Manager to manage the eLearning course assignment. Select the eLearning course order and click on the Assign credits button to remove the existing user assignment.
Note: You can only change the assigned user if they have not yet accessed the eLearning course.

Can I assign an eLearning course for analytical instruments and lab equipment to more than one person?

You can assign an eLearning course for analytical instruments and lab equipment to more than one person only if you purchased multiple quantities. The number of quantities purchased equals the number of users that can be assigned to the course.

How long do I have to assign a purchased eLearning course for analytical instruments and lab equipment to a user?

You have one year from the date of purchase to assign an eLearning course for analytical instruments and lab equipment to a user.

What if the user I am delegating an eLearning course for analytical instruments and lab equipment to does not receive the email on how to manage eLearnings?

Please verify the email address used for delegating. If you are unable to locate the email, you can access Credit Manager directly.