PEEK Fingertight Fittings
Actual product may vary
PEEK Fingertight Fittings
PEEK Fingertight Fittings
PEEK Fingertight Fittings
PEEK Fingertight Fittings
PEEK Fingertight Fittings
PEEK Fingertight Fittings
Thermo Scientific™
PEEK Fingertight Fittings
Ensure HPLC quality and robustness with Thermo Scientific™ PEEK Fingertight Fittings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeDescription
U-467BLKShort Head Port Plug, Black1/16 in., 10-32 Coned, Delrin, Black
F-300XPEEK Wing Nut w/PEEK Ferrule1/16 in.
M-645XFingertight FittingPEEK, 1/32 in., 0.25 in. Head
U-467RXColumn End Plugs, Red1/16 in., 10-32 Coned, Delrin, Red
Catalog number U-467BLK
Price (USD)
2.82
Each
Type:
Short Head Port Plug, Black
Description:
1/16 in., 10-32 Coned, Delrin, Black
Price (USD)
2.82
Each
Ensure HPLC quality and robustness with Thermo Scientific™ PEEK Fingertight Fittings. Machined for reliability, these fittings resist cracking, breaking, thread stripping and leaking in both low- and high-pressure applications. Fingertight plumbing ensures ease of use. The fittings are biocompatible for use across a broad range of applications.
Description
  • Resist cracking, breaking, thread stripping and leaking in both low and high pressure applications
  • Biocompatible for a broad range of applications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description1/16 in., 10-32 Coned, Delrin, Black
Quantity1 Each
For Use With (Equipment)10-32 Coned Port
MaterialDelrin Acetal
TypeShort Head Port Plug, Black
Unit SizeEach