Revco™ CxF Series -86°C Ultra-Low Temperature Chest Freezer, 17 cu. ft. (481.4L) capacity, 230V/50Hz, 12A - FAQs
Revco™ CxF Series -86°C Ultra-Low Temperature Chest Freezer, 17 cu. ft. (481.4L) capacity, 230V/50Hz, 12A - FAQs
View additional product information for Revco™ CxF Series -86°C Ultra-Low Temperature Chest Freezers - FAQs (ULT1790-10-V)