Unsorted TLD Materials
Actual product may vary
Thermo Scientific™
Unsorted TLD Materials
Thermo Scientific™ Unsorted TLD Materials include TLD 100, 600, and 700 materials consisting of Lithium Fluoride LiF:Ti, Mg.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Application)
UNO68835Neutron Dosimetry Applications
UNO78835Gamma and Beta Dosimetry Applications
UNO18835Health and Medical Physics Dosimetry Applications
Catalog number UNO68835
Price (USD)
-
For Use With (Application):
Neutron Dosimetry Applications
Thermo Scientific™ Unsorted TLD Materials include TLD 100, 600, and 700 materials consisting of Lithium Fluoride LiF:Ti, Mg. Suitable for health and medical physics, γ,β, and neutron dosimetry applications, depending on material type.
  • TLD-100 materials consist of Lithium Fluoride (Li natural) LiF:Mg, Ti; suitable for health and medical physics dosimetry applications
  • TLD-600 materials consist of Lithium Fluoride (Lithium isotope) Li:Mg, Ti; suitable for neutron dosimetry applications
  • TLD-700 materials consist of Lithium Fluoride (Lithium isotope) Li:Mg, Ti; suitable for gamma and beta dosimetry applications
Specifications
Accuracy30
DescriptionModel TLD- 600
For Use With (Application)Neutron Dosimetry Applications
Radiation TypeNeutron
Yield∼40 chips per gram
MaterialLithium Fluoride (Lithium isotope) Li:Mg, Ti
Product TypeThermoluminescent Dosimetry Material
Unit SizeEach