Thermoluminescent Dosimetry Powders
Actual product may vary
Thermo Scientific™

Thermoluminescent Dosimetry Powders

Get accurate radiation dose measurement for any application, from health physics to environmental dosimetry applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Application)Emission Spectra
UNW28020Environmental Dosimetry4835 Å
UCW71727Gamma, Beta and Environmental4000 Å
UNW78020Gamma and Beta Dosimetry3500 to 6000 Å (4000 maximum)
UNW68020Neutron Dosimetry3500 to 6000 Å (4000 maximum)
UCW11727Environmental, Personnel, Extremity Dosimetry4000 Å
UCW61727Neutron Dosimetry4000 Å
UNW18020Health and Medical Physics Dosimetry3500 to 6000 Å (4000 maximum)
Catalog number UNW28020
Price (MXN)
-
For Use With (Application):
Environmental Dosimetry
Emission Spectra:
4835 Å

Get accurate radiation dose evaluation for applications from health physics to environmental to personnel and extremity, and beyond with highly sensitive thermoluminescent dosimetry powders. Thermo Scientific™ Thermoluminescent Dosimetry Powders are accurate for x-ray radiation, gamma radiation, beta radiation, neutron radiation, and electron radiation according to choice of material.

TLD-600 materials consist of Lithium Fluoride (Li isotope) LiF:Mg, Ti; suitable for gamma and beta dosimetry applications.


Specifications
DescriptionModel No.TLD-200
For Use With (Application)Environmental Dosimetry
Measurement Ranges0.1 pGy to 10 Gy
Emission Spectra4835 Å
Energy Response∼12.5
MaterialCalcium Fluoride Dysporium CaF2:Dy
Product TypeThermoluminescent Dosimetry Material
Sensitivity30 at 5765å
Unit SizeEach