Revco™ UxF -86°C Upright Ultra-Low Temperature Freezer, 14.9 cu. ft. (420.9L) capacity, holds 300 boxes, 208-230V/60Hz - FAQs
Revco™ UxF -86°C Upright Ultra-Low Temperature Freezer, 14.9 cu. ft. (420.9L) capacity, holds 300 boxes, 208-230V/60Hz - FAQs
View additional product information for Revco™ UxF -86°C Upright Ultra-Low Temperature Freezers - FAQs (UXF30086D)