Revco™ UxF -86°C Upright Ultra-Low Temperature Freezer, 33.5 cu. ft. (948.7L) capacity, holds 700 boxes, 208-230V/60Hz - FAQs
Revco™ UxF -86°C Upright Ultra-Low Temperature Freezer, 33.5 cu. ft. (948.7L) capacity, holds 700 boxes, 208-230V/60Hz - FAQs
View additional product information for Revco™ UxF -86°C Upright Ultra-Low Temperature Freezers - FAQs (UXF70086D)