CytoVista™ 3D Cell Culture Clearing Reagent, 30 mL - FAQs
CytoVista™ 3D Cell Culture Clearing Reagent, 30 mL - FAQs
View additional product information for CytoVista™ 3D Cell Culture Clearing Reagent - FAQs (V11315)
4 product FAQs found
Can spheroids or organoids be imaged and stored in CytoVista 3D Cell Culture Clearing Reagents (Cat. Nos. V11315, V11326)?
CytoVista 3D Cell Culture Clearing Reagent (Cat. Nos. V11315, V11326) can be used as an imaging solution. However, we do not recommend keeping the spheroids or organoids in the clearing reagent for more than two weeks.
Is CytoVista 3D Cell Culture Clearing Reagent (Cat. No. V11315, V11326) compatible with 3D cell culture matrices like Matrigel matrix (Corning)?
No. CytoVista 3D Cell Culture Clearing Reagent (Cat. No. V11315, V11326) is incompatible with most 3D cell culture matrices. We recommend harvesting the cells from the matrix to enhance the penetration of antibodies and other staining reagents.
Do I have to use CytoVista 3D Cell Culture Clearing Reagent under a chemical safety hood?
We recommend following the usage guidelines specified in the MSDS.